Skip to Content
Merck
All Photos(2)

Key Documents

GF46548692

Vanadium

microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%

Synonym(s):

Vanadium, V 004850

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
MDL number:
UNSPSC Code:
12141749
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

8 mmHg ( 20 °C)

Assay

99.8%

form

foil

manufacturer/tradename

Goodfellow 465-486-92

resistivity

24.8-26.0 μΩ-cm, 20°C

diam. × thickness

10 mm × 1.0 μm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine Leblanc et al.
Biochimie, 88(11), 1773-1785 (2006-09-30)
Brown algal kelp species are the most efficient iodine accumulators among all living systems, with an average content of 1.0% of dry weight in Laminaria digitata, representing a ca. 30,000-fold accumulation of this element from seawater. Like other marine macroalgae
Zu Liang Chen et al.
Analytica chimica acta, 607(1), 1-14 (2007-12-25)
A comprehensive review is presented addressing recent trends in the speciation and determination of vanadium in environmental and biological sample matrices, including important analytical aspects such as sample clean up, pre-concentration and method development. Methodology based on both separation and
Anupam Bishayee et al.
Cancer letters, 294(1), 1-12 (2010-03-09)
Vanadium, a dietary micronutrient, is yet to be established as an essential part of the human diet. Over the past century, several biological effects of vanadium, such as insulin-mimetic action as well as amelioration of hyperlipidemia and hypertension, have been
D A Barrio et al.
Current medicinal chemistry, 17(31), 3632-3642 (2010-09-18)
Vanadium is a trace element present in practically all cells in plants and animals. While the essentiality of vanadium for human beings remains to be well established, vanadium has become an increasingly important environmental metal. Vanadium compounds exert a variety
Birgit Schwenzer et al.
ChemSusChem, 4(10), 1388-1406 (2011-11-22)
Large-scale energy storage has become the main bottleneck for increasing the percentage of renewable energy in our electricity grids. Redox flow batteries are considered to be among the best options for electricity storage in the megawatt range and large demonstration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service