Skip to Content
Merck
All Photos(1)

Key Documents

774073

Sigma-Aldrich

Vanadium

pieces, 1-3 mm, 99.9% trace metals basis

Synonym(s):

Vanadium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
EC Number:
MDL number:
UNSPSC Code:
12141749
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

8 mmHg ( 20 °C)

Quality Level

Assay

99.9% trace metals basis

form

pieces

resistivity

24.8-26.0 μΩ-cm, 20°C

particle size

1-3 mm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Vanadium metal is commonly used industrially as a key component in alloys[1]. Vanadium has also been shown to increase the efficiency of photocatalysis in TiO2[2] and hydrogen uptake in MgH2[3].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramzan; M.;
International Journal of Hydrogen Energy, 35(19), 10373-10376 (2010)
Anpo; M.;
Catalysis Surveys from Japan, 1(2), 169-179 (1997)
Gonzalez; J.E.G.;
Journal of Electroanalytical Chemistry (Lausanne Switzerland), 471(2), 109-115 (1999)
Dieter Rehder
Journal of inorganic biochemistry, 102(5-6), 1152-1158 (2008-02-08)
In the present focused review, vanadate-dependent haloperoxidases and vanadate-inhibited enzymes which catalyze the hydrolysis of phosphoester bonds are addressed. In these systems, vanadate [HxVO4](3-x)(-) is covalently coordinated to the imidazolyl moiety of an active site histidine, with a geometrical arrangement
Jaclyn M Winter et al.
The Journal of biological chemistry, 284(28), 18577-18581 (2009-04-14)
Nature has developed an exquisite array of methods to introduce halogen atoms into organic compounds. Most of these enzymes are oxidative and require either hydrogen peroxide or molecular oxygen as a cosubstrate to generate a reactive halogen atom for catalysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service