Skip to Content
Merck
All Photos(1)

Key Documents

CDS000001

Sigma-Aldrich

Magnesium citrate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6MgO7
CAS Number:
Molecular Weight:
214.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

OC(=O)CC1(O)CC(=O)O[Mg]OC1=O

InChI

1S/C6H8O7.Mg/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);/q;+2/p-2

InChI key

DIXGJWCZQHXZNR-UHFFFAOYSA-L

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nusrat Hussain et al.
The Biochemical journal, 477(8), 1373-1389 (2020-03-28)
We investigated acute effects of two allosteric protein kinase B (PKB) inhibitors, MK-2206 and Akti-1/2, on insulin-stimulated lipogenesis in rat epididymal adipocytes incubated with fructose as carbohydrate substrate. In parallel, the phosphorylation state of lipogenic enzymes in adipocytes and incubated
Laura Blancquaert et al.
Nutrients, 11(7) (2019-07-25)
Despite the presumption of the beneficial effects of magnesium supplementation, little is known about the pharmacokinetics of different magnesium formulations. We aimed to investigate the value of two in vitro approaches to predict bioavailability of magnesium and to validate this
Euan Watt et al.
Microbiome, 4(1), 61-61 (2016-11-26)
Sequencing-based analysis has become a well-established approach to deciphering the composition of the gut microbiota. However, due to the complexity of accessing sufficient material from colonoscopic biopsy samples, most studies have focused on faecal microbiota analysis, even though it is
Rajaratnam Rameshshanker et al.
Gastrointestinal endoscopy, 91(4), 894-904 (2019-12-15)
Adenoma miss rate during colonoscopy is directly linked to risk of postcolonoscopy colorectal cancer. One of the reasons for missed adenomas is poor visualization of proximal folds during standard colonoscopy withdrawal. Disposable distal attachments such as the plastic cap and
S Yamamoto et al.
Biomedical imaging and intervention journal, 5(3), e28-e28 (2009-07-01)
The prevalence of colon cancer has seen strong demand in screening for colorectal neoplasia, and this has drawn considerable attention to the technological advances in Computed Tomographic Colonography (CTC). With the assistance of an oral contrast agent, an imaging technique

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service