Skip to Content
Merck
All Photos(1)

Key Documents

PHR1681

Supelco

Benzalkonium Chloride solution

50%, Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Benzalkonium chloride solution, Alkylbenzyldimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Product Name

Benzalkonium Chloride 50% Solution, Pharmaceutical Secondary Standard; Certified Reference Material

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001060
traceable to USP 1050993

API family

benzalkonium chloride

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-30°C

SMILES string

[Cl-].[N+](CCCCCCCCCC)(Cc1ccccc1)(C)C

InChI

1S/C19H34N.ClH/c1-4-5-6-7-8-9-10-14-17-20(2,3)18-19-15-12-11-13-16-19;/h11-13,15-16H,4-10,14,17-18H2,1-3H3;1H/q+1;/p-1

InChI key

TTZLKXKJIMOHHG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2095 in the slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid determination of total benzalkonium chloride content in ophthalmic formulation.
Labranche P-L, et al.
Journal of Pharmaceutical and Biomedical Analysis, 43(3), 989-993 (2007)
CRC Handbook of Food, Drug, and Cosmetic Excipients (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service