Skip to Content
Merck
All Photos(3)

Key Documents

D76609

Sigma-Aldrich

Dicyandiamide

99%

Synonym(s):

DCD, Cyanoguanidine, Dicyanodiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NHCN
CAS Number:
Molecular Weight:
84.08
Beilstein:
605637
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

208-211 °C (lit.)

SMILES string

NC(=N)NC#N

InChI

1S/C2H4N4/c3-1-6-2(4)5/h(H4,4,5,6)

InChI key

QGBSISYHAICWAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dicyandiamide is commonly used for the curing of epoxy resins. It is a nitrification inhibitor that is said to be capable of reducing nitrate (NO3-) leaching and nitrous oxide (N2O) emissions from grazed pasture soils.

Application

Dicyandiamide can be used as an organic precursor for synthesizing carbon nitride nanosheets.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafał Olchowski et al.
Materials (Basel, Switzerland), 13(7) (2020-04-05)
Ordered mesoporous carbon (CMK-3), obtained from an abundant natural source, sugar, was thermochemically modified with dicyandiamide and thiourea as well as by classical oxidization with hydrogen peroxide to introduce specific surface groups. Thermochemical modifications resulted in carbon with almost unchanged
The use of a nitrification inhibitor, dicyandiamide (DCD), to decrease nitrate leaching and nitrous oxide emissions in a simulated grazed and irrigated grassland.
Di HJ
Soil use and management, 18(4), 395-403 (2002)
F Al-Rimawi
Talanta, 79(5), 1368-1371 (2009-07-29)
A simple, and stability-indicating liquid chromatographic method was developed and validated for the analysis of metformin hydrochloride and its related compound (1-cyanoguanidine) in tablet formulations. Liquid chromatography with a UV detector at a wavelength of 232 nm using a Nova-Pak
Enrico Colli et al.
Expert opinion on investigational drugs, 16(7), 999-1007 (2007-06-28)
'Overactive bladder' (OAB) is a syndrome that is characterised by symptoms of urgency, with or without urge urinary incontinence, usually with frequency and nocturia [1] . It is a highly prevalent condition affecting 17% of the general population, with a
Ian Levett et al.
Journal of agricultural and food chemistry, 67(9), 2449-2458 (2019-02-07)
Dicyandiamide (DCD) has been studied as a stabilizer for nitrogen fertilizers for over 50 years. Its efficacy is limited at elevated temperatures, but this could be addressed by encapsulation to delay exposure. Here, poly(3-hydroxybutyrate- co-3-hydroxyvalerate) (PHBV) was investigated as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service