Skip to Content
Merck
All Photos(2)

Documents

225819

Sigma-Aldrich

Hydrazine hydrate

reagent grade, N2H4 50-60 %

Synonym(s):

Hydrazinium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · xH2O
CAS Number:
Molecular Weight:
32.05 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

>1 (vs air)

vapor pressure

5 mmHg ( 25 °C)

form

liquid

composition

Degree of hydration, ~1.5
N2H4, 50-60%

expl. lim.

99.99 %

concentration

50.0-60.0% (%H4N2 by Na2S2O3 basis, titration)

density

1.029 g/mL at 25 °C (lit.)

SMILES string

NN.[H]O[H]

InChI

1S/H4N2.H2O/c1-2;/h1-2H2;1H2

InChI key

IKDUDTNKRLTJSI-UHFFFAOYSA-N

General description

The addition of hydrazine hydrate to reduced graphene oxide (RGO) counter electrode improves its performance in dye-sensitized solar cells (DSSC).

Application

Hydrazine hydrate has been used for the deproteination of the enamel samples in a study. It may be used as a reducing agent in the following:
  • Preparation of silver nanoparticles.
  • Transformation of monosubstituted nitrobenzene derivatives to the corresponding anilines.
  • Along with graphite for the conversion of nitro compounds (aromatic and aliphatic) to the amino compounds.
Hydrazine hydrate may be used to prepare:
  • 3-(2-Benzyloxy-6-hydroxyphenyl)-5-styrylpyrazoles by reacting with 5-benzyloxy-2-styrylchromones.
  • 3,5-Diphenyl-2-pyrazoline derivatives by reacting with 1,3-diphenyl-2-propen-1-one.
  • 3′-Aryl-1,2,3,4,4′,5′-hexahydrospiro[quinoxalin-2,5′-pyrazol]-3-ones by reacting with 3-arylacylidene-3,4-dihydroquinoxalin-2(1H)-ones.
  • It may also be used in the catalytic reduction of nitroarenes to aromatic amines.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Baumann et al.
Scientific reports, 5, 15194-15194 (2015-10-16)
Enamel proteins form a scaffold for growing hydroxyapatite crystals during enamel formation. They are then almost completely degraded during enamel maturation, resulting in a protein content of only 1% (w/v) in mature enamel. Nevertheless, this small amount of remaining proteins
Graphite catalyzed reduction of aromatic and aliphatic nitro compounds with hydrazine hydrate.
Byung HH, et al.
Tetrahedron Letters, 26(50), 6233-6234 (1985)
Reduction of aromatic nitro compounds with hydrazine hydrate in the presence of an iron oxide hydroxide catalyst. I. The reduction of monosubstituted nitrobenzenes with hydrazine hydrate in the presence of ferrihydrite.
Lauwiner M, et al.
Applied Catalysis A: General, 172(1), 141-148 (1998)
Catalytic reduction of nitroarenes with hydrazine hydrate in suitable solvents.
Ayyangar NR, et al.
Synthesis, 1981(08), 640-643 (1981)
Synthesis and molecular structure of 3-(2-benzyloxy-6-hydroxyphenyl)-5-styrylpyrazoles. Reaction of 2-styrylchromones and hydrazine hydrate.
Pinto DC, et al.
Tetrahedron, 55(33), 10187-10200 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service