Skip to Content
Merck
All Photos(3)

Key Documents

C8518

Sigma-Aldrich

Cesium hydroxide hydrate

≥90%, ≥99.5% (metals basis)

Synonym(s):

Cesium hydroxide monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCsO · xH2O
CAS Number:
Molecular Weight:
149.91 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder, crystals or chunks

grade

for analytical purposes

Assay

≥90%
≥99.5% (metals basis)

form

powder, crystals or chunks

impurities

≤5% Cs2CO3

mp

272 °C (lit.)

density

3.68 g/mL at 25 °C (lit.)

SMILES string

O.[OH-].[Cs+]

InChI

1S/Cs.2H2O/h;2*1H2/q+1;;/p-1

InChI key

ABSOMGPQFXJESQ-UHFFFAOYSA-M

Application

Cesium hydroxide hydrate (CsOH.H2O) can be used in the preparation of lithium substituted cesium and cesium aluminosilicate based geopolymer as potential refractory materials. It can also be used in the synthesis of cesium structures of 1,8-naphthalimides substituted carboxylic ligands.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

XRD Analysis of the Role of Cesium in Sodium-Based Geopolymer.
Chlique C, et al.
Journal of the American Ceramic Society. American Ceramic Society, 98(4), 1308-1313 (2015)
Thermal evolution of lithium ion substituted cesium-based geopolymer under high temperature treatment, Part I: Effects of holding temperature
Yuan J, et al.
Ceramics International, 44(9), 10047-10054 (2018)
Cesium hydroxide catalyzed addition of alcohols and amine derivatives to alkynes and styrene
Dimitrios T, et al.
Tetrahedron Letters, 40(34), 6193-6195 (1999)
Atomic structure of a cesium aluminosilicate geopolymer: a pair distribution function study.
Bell J, et al.
Chemistry of Materials, 20(14), 4768-4776 (2008)
Cesium complexes of naphthalimide substituted carboxylate ligands: Unusual geometries and extensive cation-$\pi$ interactions
Reger DL, et al.
Journal of Molecular Structure, 1091(14), 31-36 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service