Skip to Content
Merck
All Photos(1)

Documents

C8136

Sigma-Aldrich

Chloranilic acid

≥98%

Synonym(s):

2,5-Dichloro-3,6-dihydroxy-2,5-cyclohexadiene-1,4-dione, 2,5-Dichloro-3,6-dihydroxy-p-benzoquinone, NSC 6108, NSC 97383

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl2O4
CAS Number:
Molecular Weight:
208.98
Beilstein:
1875040
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

≥300 °C (lit.)

SMILES string

OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O

InChI

1S/C6H2Cl2O4/c7-1-3(9)5(11)2(8)6(12)4(1)10/h9,12H

InChI key

IPPWILKGXFOXHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Acting as a proton donor in reactions studying dimensionality control
  • Synthesis of dimethylbipyridyl complexes
  • Synthesis of (nonylbenzimidazolylmethyl)benzene for preparation of neutral altitudinal rotor-shaped dirhenium metallacycles
  • Charge-transfer reactions with metoprolol tartrate
  • Salt formation with organic bases
  • Synthesis of osmium metalla-rectangles with anticancer activity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis G Treviño-Quintanilla et al.
Journal of microbiology (Seoul, Korea), 49(6), 974-980 (2011-12-29)
Pentachlorophenol is the most toxic and recalcitrant chlorophenol because both aspects are directly proportional to the halogenation degree. Biological and abiotic pentachlorophenol degradation generates p-chloranil, which in neutral to lightly alkaline environmental conditions is hydrolyzed to chloranilic acid that present
M Walash et al.
Archives of pharmacal research, 27(7), 720-726 (2004-09-11)
Two Spectrophotometric procedures are presented for the determination of two commonly used H2-receptor antagonists, nizatidine (I) and ranitidine hydrochloride (II). The methods are based mainly on charge transfer complexation reaction of these drugs with either p-chloranilic acid (rho-CA) or 2
Yasuhiro Tobu et al.
Physical chemistry chemical physics : PCCP, 14(35), 12347-12354 (2012-08-09)
The temperature dependence of (35)Cl NQR frequencies and the spin-lattice relaxation times T(1) has been measured in the wide temperature range of 4.2-420 K for morpholinium hydrogen chloranilate in which a one-dimensional O-HO hydrogen-bonded molecular chain of hydrogen chloranilate ions
Said M Teleb et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(1-3), 140-145 (2005-11-01)
Charge-transfer (CT) complexes formed on the reaction of 2,2'-bipyridine with some acceptors such as picric acid (HPA) and chloranilic acid (H(2)CA) have been studied in CHCl(3) and MeOH at room temperature. Based on elemental analysis and IR spectra of the
Elmorsy Khaled
Talanta, 75(5), 1167-1174 (2008-07-01)
A simple, rapid and accurate method for the spectrophotometric determination of terfenadine has been developed. The proposed method based on the charge-transfer reactions of terfenadine, as n-electron donor, with 7,7,8,8-tetracyanoquinodimethane (TCNQ), tetracyanoethylene (TCNE), 2,3-dichloro-5,6-dicyano-1,4-benzoquinone (DDQ) or 2,5-dichloro-3,6-dihydroxy-1,4-benzoquinone (chloranilic acid, p-CLA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service