128562
Hexaketocyclohexane octahydrate
97%
Synonym(s):
Cyclohexanehexone octahydrate, Triquinoyl octahydrate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6O6 · 8H2O
CAS Number:
Molecular Weight:
312.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
powder
mp
99 °C (dec.) (lit.)
functional group
ketone
storage temp.
2-8°C
SMILES string
[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].O=C1C(=O)C(=O)C(=O)C(=O)C1=O
InChI
1S/C6O6.8H2O/c7-1-2(8)4(10)6(12)5(11)3(1)9;;;;;;;;/h;8*1H2
InChI key
MQIMWEBORAIJPP-UHFFFAOYSA-N
General description
Hexaketocyclohexane octahydrate also known as Triquinoyl octahydrate is an organic compound, often utilized as a building block in various organic reactions, including the synthesis of water-soluble π-conjugated hexaazatrinaphthylenes and their derivatives.
Application
Hexaketocyclohexane octahydrate has been used in the preparation of hexaazatriphenylenehexacarbonitrile.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and some reactions of hexaazatriphenylenehexanitrile, a hydrogen-free polyfunctional heterocycle with D3h symmetry.
Kanakarajan K and Czarnik AW.
The Journal of Organic Chemistry, 51(26), 5241-5243 (1986)
Hao Huang et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(9), 2000012-2000012 (2020-05-10)
The 2D conductive metal-organic frameworks (MOFs) are expected to be an ideal electrocatalyst due to their high utilization of metal atoms. Exploring a new conjugated ligand with extra active metallic center can further boost the structural advantages of conductive MOFs.
Lei Wang et al.
Nanoscale, 9(12), 4090-4096 (2017-03-16)
Nanostructured semiconducting polymers have emerged as a very promising class of metal-free photocatalytic materials for solar water splitting. However, they generally exhibit low efficiency and lack the ability to utilize long-wavelength photons in a photocatalytic oxygen evolution reaction (OER). Here
Nitrogen-rich layered carbon for adsorption of typical volatile organic compounds and low-temperature thermal regeneration
Li Tan, et al.
Journal of Hazardous Materials, 424, 127348-127348 (2022)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service