Skip to Content
Merck
All Photos(3)

Key Documents

542040

Sigma-Aldrich

Potassium nitrate

99.999% trace metals basis

Synonym(s):

Saltpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
crystalline

Quality Level

Assay

99.999% trace metals basis

form

crystalline

technique(s)

flow cytometry: suitable

impurities

≤15.0 ppm Trace Metal Analysis

mp

334 °C (lit.)

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Improving drought and salinity tolerance in barley by application of salicylic acid and potassium nitrate: Studies the ameliorative effects of potassium nitrate and salicylic acid on barley, showing improved tolerance to drought and salinity stress (KA Fayez, SA Bazaid, 2014).
  • Prior application of 10% potassium nitrate to reduce post-bleaching sensitivity: a randomized triple-blind clinical trial: Evaluates the use of potassium nitrate in reducing sensitivity post dental bleaching, providing clinical evidence of its effectiveness (M Rezende, KL da Silva, TC Miguel, PV Farago, 2020).

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rose A Devasia et al.
The Journal of antimicrobial chemotherapy, 63(6), 1173-1178 (2009-03-31)
The aim of this study was to assess the sensitivity, specificity and time to results of mycobacterial growth indicator tube (MGIT) 960, microscopic observation drug susceptibility (MODS) assay and nitrate reductase assay (NRA) compared with the gold standard agar proportion
Jing Huang et al.
Applied and environmental microbiology, 80(1), 29-42 (2013-10-01)
Bacteria play important roles in mineral weathering and soil formation. However, few reports of mineral weathering bacteria inhabiting subsurfaces of soil profiles have been published, raising the question of whether the subsurface weathering bacteria are fundamentally distinct from those in
Changhong Yao et al.
Bioresource technology, 118, 438-444 (2012-06-22)
Microalgal starch is a potential feedstock for biofuel production. The effects of KNO(3) and MgSO(4) concentrations and light intensity on biomass and starch production by the marine microalga, Tetraselmis subcordiformis, were investigated. Under 200 μmol m(-2) s(-1) irradiance and sulfur-deprived
Yu Yang et al.
Environmental science & technology, 48(11), 6391-6400 (2014-04-24)
Titanium dioxide (TiO2) is widely used in food products, which will eventually enter wastewater treatment plants and terrestrial or aquatic environments, yet little is known about the fraction of this TiO2 that is nanoscale, or the physical and chemical properties
Oliver Peacock et al.
Medicine and science in sports and exercise, 44(11), 2213-2219 (2012-08-10)
The objective of this study is to examine the effects of acute ingestion of dietary nitrate on endurance running performance in highly trained cross-country skiers. Dietary nitrate has been shown to reduce the oxygen cost of submaximal exercise and improve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service