Skip to Content
Merck
All Photos(1)

Documents

B32602

Sigma-Aldrich

Benzyltrimethylammonium hydroxide solution

40 wt. % in methanol

Synonym(s):

N,N,N-Trimethyl-N-benzylammonium hydroxide, N,N,N-trimethyl-1-phenylmethanaminium hydroxide, N,N,N-trimethylbenzenemethanaminium hydroxide, Triton B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(OH)(CH3)3
CAS Number:
Molecular Weight:
167.25
Beilstein:
3917256
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

40 wt. % in methanol

density

0.92 g/mL at 25 °C

SMILES string

[OH-].C[N+](C)(C)Cc1ccccc1

InChI

1S/C10H16N.H2O/c1-11(2,3)9-10-7-5-4-6-8-10;/h4-8H,9H2,1-3H3;1H2/q+1;/p-1

InChI key

NDKBVBUGCNGSJJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Benzyltrimethylammonium hydroxide solution is a quaternary ammonium compound that is commonly used as a base in organic synthesis. It is also used as a base catalyst in some polymerization reactions.

Application

Benzyltrimethylammonium hydroxide solution can be used as:     
  • A catalyst in the nitroaldol condensation reaction      
  • A structure-directing agent in the synthesis of high-silica aluminosilicate zeolite chabazite type zeolite by hydrothermal method      
  • A ionic liquid precursor for the fabrication of nanostructured ZnO particles

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Sangeetha Vedula et al.
Archives of biochemistry and biophysics, 466(2), 260-266 (2007-08-07)
Trichodiene synthase is a terpenoid cyclase that catalyzes the cyclization of farnesyl diphosphate (FPP) to form the bicyclic sesquiterpene hydrocarbon trichodiene (89%), at least five sesquiterpene side products (11%), and inorganic pyrophosphate (PP(i)). Incubation of trichodiene synthase with 2-fluorofarnesyl diphosphate
Lena Edström et al.
Journal of chromatography. A, 1218(15), 1966-1973 (2010-10-05)
It has recently been demonstrated, using mathematical models, how peculiar overloaded band profiles of basic compounds are due to the local pH in the column when using low capacity buffers. In this study, overloaded peak shapes resulting after injection of
Weisheng Lin et al.
Analytica chimica acta, 583(1), 98-102 (2007-03-28)
The importance of benzyltriethyl ammonium chloride (BTEAC) in industrial applications has stimulated the development of a number of methods for its determination. In this paper, a high performance capillary electrophoresis (CE) method, coupled with an extraction technique for determining BTEAC
Purushothaman Gopinath et al.
The Journal of organic chemistry, 74(16), 6291-6294 (2009-07-22)
An efficient protocol is reported for the synthesis of thioesters from carboxylic acids with use of acyloxy phosphonium salts as intermediates and benzyltriethylammonium tetrathiomolybdate as the sulfur transfer reagent.
Nicola H Davies et al.
Journal of chromatography. A, 1119(1-2), 11-19 (2006-01-03)
The retention and overloading properties for eight basic solutes and two quaternary ammonium compounds were studied over the pH range 2.7-10.0 using phosphate and carbonate buffers. At low pH, a hybrid inorganic-organic silica-ODS phase (XTerra RP-18, 15 cm x 0.46

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service