Skip to Content
Merck
All Photos(3)

Key Documents

327182

Sigma-Aldrich

tert-Butyl acrylate

contains 10-20 ppm monomethyl ether hydroquinone as inhibitor, 98%

Synonym(s):

t-Butyl acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOC(CH3)3
CAS Number:
Molecular Weight:
128.17
Beilstein:
1742329
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

contains

10-20 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.410 (lit.)

bp

61-63 °C/60 mmHg (lit.)

density

0.875 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OC(=O)C=C

InChI

1S/C7H12O2/c1-5-6(8)9-7(2,3)4/h5H,1H2,2-4H3

InChI key

ISXSCDLOGDJUNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl acrylate is an organic chemical compound that can be used as a monomer for the preparation of various types of polymers with desirable characteristics. It can undergo free-radical polymerization, which is a common method used to produce polymeric materials. Additionally, tert-butyl acrylate polymers can be copolymerized with other monomers to modify their properties. The versatility of tert-butyl acrylate makes it suitable for a wide range of applications, including coatings, adhesives, sealants, elastomers, and specialty films.

Application

tert-Butyl acrylate is a monomer used to produce poly(tert-butyl acrylate) (PtBA) polymers and copolymers, and poly(acrylic acid) polyelectrolyte brushes. It undergoes radiation polymerization to form TBA-gels for radio-fluorogenic dose imaging applications. The monomer can be used to synthesize Poly(t-butyl acrylate). It may be used as a reactive diluent in a study.

tert-Butyl acrylate can also be used as:
  • A building block in the synthesis of asymmetric membranes, which are potentially useful in filtration, separation, or gas permeation processes.
  • A monomer in the preparation of a siloxane-incorporated copolymer binder for silicon anodes in lithium-ion batteries.
  • A monomer in the synthesis of poly(tert-butyl acrylate)/carbon nanofiber (CNF) materials through atom transfer radical polymerization (ATRP).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymer-gel formation and reformation on irradiation of tertiary-butyl acrylate
Yao T, et al.
Radiation Physics and Chemistry, 97, 147-152 (2014)
Huanhuan Liu et al.
Polymers, 11(5) (2019-05-10)
Gambogenic acid (GNA) has been demonstrated with outstanding antitumor activity as a potential antitumor drug in recent years. However, the low solubility and deficient bioavailability of GNA seriously hinder its practical application in the clinic area. In this study, a
Study of the kinetics of step and flash imprint lithography photopolymerization.
Dickey MD, et al.
AIChE Journal, 51(9), 2547-2555 (2005)
Daniel Hassan et al.
Pharmaceutics, 12(11) (2020-11-19)
Globally, human beings continue to be at high risk of infectious diseases caused by methicillin-resistant Staphylococcus aureus (MRSA); and current treatments are being depleted due to antimicrobial resistance. Therefore, the synthesis and formulation of novel materials is essential for combating
Calvin A Omolo et al.
Journal of liposome research, 31(1), 45-63 (2019-10-31)
pH responsive drug delivery systems are one of the new strategies to address the spread of bacterial resistance to currently used antibiotics. The aim of this study was to formulate liposomes with 'On' and 'Off'' pH responsive switches for infection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service