Skip to Content
Merck
All Photos(1)

Key Documents

925950

Sigma-Aldrich

Titanium Aluminum Carbonitride MAX phase

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti3AlCN
Molecular Weight:
196.60
UNSPSC Code:
12352311
NACRES:
NA.23

form

powder

Quality Level

color

gray to black

solubility

insoluble

SMILES string

[Ti][Ti][Ti][Al]([N])[C]

Application

Titanium Aluminum Carbonitride MAX phase is an excellent MXene precursor; and can be processed by HF or HCl+LiF to obtain MXene.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service