Recommended Products
Application
Titanium Aluminum Nitride MAX phase has low density, high elastic modulus, easy machinability, and excellent thermal shock resistance. This product is an excellent MXene precursor, and can be processed by HF or HCl+LiF to obtain MXene.
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and microstructure of layered-ternary Ti2AlN ceramic
Scr. Mater., 56, 1115?1118-1115?1118 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service