Skip to Content
Merck
All Photos(1)

Key Documents

921408

Sigma-Aldrich

Titanium Carbide MXene dispersion in propylene carbonate

Synonym(s):

Titanium Carbide MXene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti3C2Tx
CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

form

dispersion

Quality Level

concentration

≥0.5 mg/mL in propylene carbonate

color

faint gray to black

application(s)

battery manufacturing

storage temp.

−20°C

General description

This high purity Ti3C2Tx product is a two-dimentional (2D) titanium carbide (Ti3C2) MXene material dispersed into propylene carbonate at a concentration of 1 mg/mL, where Tx is Cl, F, and OH. This product is packaged under an inert atmosphere giving it longer shelf life compared to water based dispersions. It may contain traces of Ti2AlC.

Application

This MXene (Ti3C2) dispersion product contains delaminated MXene flakes suspended in Propylene carbonate provide a stable suspension versus aqueous-based dispersions. Propylene carbonate between the high surface area MXene layers may form a continuous interface between the MXene layers and the PC/EC electrolyte used in the Na-ion battery, possibly allowing easier diffusion of Na+ into the electrodes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dispersions of Two-Dimensional Titanium Carbide MXene in Organic Solvents.
Chemistry of Materials, 29(4), 1632-1640 (2017)
Highly conductive optical quality solution-processed films of 2D titanium carbide.
Dillon A D, et al.
Advances in Functional Materials, 26, 4162-4168 (2016)
2D Ti3C2Tz MXene Synthesized by Water-free Etching of Ti3AlC2 in Polar Organic Solvents.
Natu V, et al.
Chem, 6(3), 616-630 (2020)
Michael Naguib et al.
ACS nano, 6(2), 1322-1331 (2012-01-28)
Herein we report on the synthesis of two-dimensional transition metal carbides and carbonitrides by immersing select MAX phase powders in hydrofluoric acid, HF. The MAX phases represent a large (>60 members) family of ternary, layered, machinable transition metal carbides, nitrides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service