Skip to Content
Merck
All Photos(1)

Key Documents

924962

Sigma-Aldrich

Titanium Carbide (Ti3C2Tx)

Few layer MXene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti3C2Tx
UNSPSC Code:
12352103
NACRES:
NA.23

description

Electrical conductivity: 106 S/m (MXene-only film prepared from an aqueous dispersion of MXene, at RT via a four-probe technique)

Quality Level

L

1-4 μm , average lateral size (by scanning electron microscopy)

surface area

~146 m2/g , Brunauer–Emmett–Teller method

thickness

1-3 nm , by atomic force microscopy

solubility

H2O: ≥300 mg/mL

density

4.12 g/cm3 (measured by Helium pycnometer)

storage temp.

2-8°C

General description

Mxenes find extensive use in LiBs and rechargeable non-lithium-ion (Na+, K+, Mg2+, Ca2+, and Al3+) batteries. This MXene is a suitable candidate for Li-ion batteries because of its higher electrical conductivity and Li storage capacity comparable with graphite electrodes. This MXene has been explored as anode , cathode and separators in battery applications.

Application

Ti3C2Tx MXenes are a novel type of two-dimensional (2D) material composed of layered carbides of transition metals. MXenes have excellent thermal and electrical conductivity, heat resistance, and excellent volumetric capacitance, whilst having the highest electromagnetic (EMI) shielding effectiveness among similar synthetic 2D materials. Among its several applications, MXenes have been highlighted for:
  • Active material for energy storage devices (e.g. supercapacitors, lithium-based batteries);
  • Nanocomposites for power generation;
  • Biomedical applications;
  • Catalyst;
  • EMI Shielding.

Legal Information

For research only

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MXenes for advanced separator in rechargeable batteries
An, Yongling et. al.
Materials Today, 57, 146-179 (2017)
Flexible MXene/Graphene Films for Ultrafast Supercapacitors with Outstanding Volumetric Capacitance.
Yan J, et al.
Advanced Functional Materials , 27, 1701264-1701264 (2017)
Prediction and Characterization of MXene Nanosheet Anodes for Non-Lithium-Ion Batteries
Xie, Yu et. al.
ACS Nano, 8, 9606?9615-9606?9615 (2014)
Single platinum atoms immobilized on an MXene as an efficient catalyst for the hydrogen evolution reaction.
Zhang J, et al.
Nature catalysis, 1, 985-992 (2018)
MXene as a novel intercalation-type pseudocapacitive cathode and anode for capacitive deionization
Srimuk, Pattarachai et. al.
Journal of Material Chemistry A, 4, 18265-18271 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service