Skip to Content
Merck
All Photos(2)

Key Documents

920320

Sigma-Aldrich

Lithium acetate

greener alternative

anhydrous, 99.9% trace metals basis

Synonym(s):

″Acetic acid, lithium salt″, Lithium ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiOOCCH3
CAS Number:
Molecular Weight:
65.99
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Assay:
99.9% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.9% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

283-285 °C (lit.)

greener alternative category

SMILES string

[Li+].CC([O-])=O

InChI

1S/C2H4O2.Li/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

XIXADJRWDQXREU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithiumacetate (LiOAc), a lithium salt derived from acetic acid, possesses moderate water solubility, and exhibits a relatively high ionic conductivity. These properties make it useful in various electrochemical applications. It is also used as a catalyst in certain chemical reactions and as a reagent for the synthesis of lithium-based compounds.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium acetate can be used:
  • As an additive in the formation of a "water-in-bisalt" electrolyte for the application in rechargeable lithium batteries. It helps to regulate the cation size and composition of the electrolyte, influencing its conductivity, stability, and overall electrochemical properties.
  • As a component in the fabrication of solid polymer electrolytes. It enhances the ionic conductivity and electrochemical properties of the polymer electrolyte for its application as a separator in lithium-ion batteries.
  • As a component in the development of nanocomposite polymer electrolytes for solid-state supercapacitor applications.


Lithium acetate is used to create lithium containing perovskite thin films. Additionally, lithium ethanoate can be utilized to create thin film electrodes for lithium ion batteries, such as spinel Li4Ti5O12.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of La0.5Li0.5TiO3 perovskite thin films by the sol-gel method.
Kitaoka K, et al
J. Mater. Sci., 32(8), 2063-2070 (1997)
Sol−gel preparation and characterization of perovskite lanthanum lithium titanate.
Popovici C,et al.
J. Mater. Sci., 42, 3373-3377 (2007)
Nanocrystalline LiMn2O4 thin film cathode material prepared by polymer spray pyrolysis method for Li-ion battery
Karthick S N, et al.
J. Alloy Compounds, 489(2), 674-677 (2010)
Li4Ti5O12 thin-film electrodes by sol?gel for lithium-ion microbatteries.
Mosa J, et al.
Journal of Power Sources, 244(15), 482-487 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service