Skip to Content
Merck
All Photos(3)

Key Documents

517992

Sigma-Aldrich

Lithium acetate

greener alternative

99.95% trace metals basis

Synonym(s):

Lithium ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiOOCCH3
CAS Number:
Molecular Weight:
65.99
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95% trace metals basis

form

powder or crystals

reaction suitability

core: lithium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

283-285 °C (lit.)

greener alternative category

SMILES string

[Li+].CC([O-])=O

InChI

1S/C2H4O2.Li/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

XIXADJRWDQXREU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium acetate 99.95% trace metals basis with minimal contaminant levels (< 550.0 ppm) make it an ideal reagent for processes requiring stringent purity standards, contributing to advancements in research and industrial applications.

Application

Lithium acetate is a very common salt of great importance due to its many and varied uses. Lithium acetate is used to prepare buffer solutions, which are essential for various analytical procedures, including electrophoresis and chromatography. [1] It can be used as an additive in electrolytes for lithium-ion batteries, contributing to the performance and stability of the batteries. Also it is suitable to use in thin-film deposition processes for the production of electronic components, ensuring high purity and minimal contamination. [2] Lithium acetate can be used as a catalyst or a catalyst support in various chemical reactions, enhancing reaction efficiency and product yield. In a study it has been found that Lithium acetate-catalyzed aldol reaction between trimethylsilyl enolates and aldehydes proceeded smoothly in a DMF or pyridine solvent to afford the corresponding aldols in good to high yields under weakly basic conditions. [3] In general, the product is designed to support R&D activities for critical applications in analytical chemistry, biotechnology, pharmaceuticals, materials science, catalysis, electronics, and environmental testing.

Features and Benefits

As a trace metal grade reagent, lithium acetate ensures minimal interference from contaminants in trace metal analysis, providing high-purity results.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Visualization of the synergistic effect of lithium acetate and single-stranded carrier DNA on Saccharomyces cerevisiae transformation
Pham TA, et al.
Current Genetics, 57(4), 233-239 (2011)
Effect of oleic acid plasticizer on chitosan-lithium acetate solid polymer electrolytes
Yahya MZA and Arof AK
European Polymer Journal, 39(5), 897-902 (2003)
Sunhyung Park et al.
Food microbiology, 62, 202-206 (2016-11-28)
The aim of this study was to investigate the lethal effects of simultaneous treatments with gaseous chlorine dioxide (ClO2) and mild wet heat (55 °C at 100% relative humidity [RH]) on Salmonella enterica in chicken feces on the surface of eggshells.
Studies on lithium acetate doped chitosan conducting polymer system
Yahya MZA and Arof AK
European Polymer Journal, 38(6), 1191-1197 (2002)
Nirbhay Kumar Kushwaha et al.
PLoS pathogens, 13(8), e1006587-e1006587 (2017-09-01)
Geminiviruses constitute a group of plant viruses, with a ssDNA genome, whose replication in the nucleus of an infected cell requires the function of geminivirus-encoded replication initiator protein (Rep). Our results suggest that monoubiquitinated histone 2B (H2B-ub) promotes tri-methylation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service