Skip to Content
Merck
All Photos(3)

Documents

202444

Sigma-Aldrich

Poly(ethylene glycol)

average MN 3,350, hydroxyl, powder

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

product name

Poly(ethylene glycol), average Mn 3,350, powder

form

powder

mol wt

average Mn 3,350

pH

4.5-7.5

viscosity

90 cSt(210 °F) (99 °C)(lit.)

mp

54-58 °C (lit.)

density

1.204 g/mL at 25 °C

Ω-end

hydroxyl

α-end

hydroxyl

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a biocompatible inert synthetic polymer with repeating units of ether oxygen (-CH2-CH2-O-). It can be synthesized via anionic polymerization of ethylene oxide. It is extensively used in biological and medical applications due to its non-immunogenicity and high water solubility.

Application

PEG can be used to functionalize magnetite nanoparticles. The coating of the magnetic core with PEG provides good stabilization while maintaining the magnetic properties of the nanoparticles.

It can be used to modify the surface of photocatalytic TiO2 nanopowder for its application in self-cleaning paints. PEG-coated TiO2 nanopowder can be prepared under mild conditions and show excellent colloidal stability.

It can be used as a cross-linker to prepare poly(N-isopropylacrylamide) based thermosensitive injectable hydrogels. The addition of PEG improves the chemical and mechanical properties of hydrogel and prevents it from dissolving in the swelling medium. Owing to their biocompatibility and biodegradability, these hydrogels are widely used for biomedical applications.

Features and Benefits

  • High structural flexibility
  • Biocompatibility
  • High hydration capacity
  • Devoid of any steric hindrance

Other Notes

Molecular weight: Mn 3,015-3,685

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grant A Robinson et al.
Journal of neuroscience research, 94(7), 636-644 (2016-03-21)
Functional recovery following a peripheral nerve injury is made easier when regenerating axons correctly reinnervate their original targets. Polyethylene glycol (PEG) has recently been used in attempts to fuse severed peripheral axons during suture-based repair, but an analysis of target
Erinna F Lee et al.
Autophagy, 15(5), 785-795 (2019-01-11)
BECN1/Beclin 1 is a critical protein in the initiation of autophagosome formation. Recent studies have shown that phosphorylation of BECN1 by STK4/MST1 at threonine 108 (T108) within its BH3 domain blocks macroautophagy/autophagy by increasing BECN1 affinity for its negative regulators
Yixu Wang et al.
mSphere, 4(3) (2019-05-31)
Sporisorium scitamineum is the fungal pathogen causing severe sugarcane smut disease that leads to massive economic losses globally. S. scitamineum invades host cane by dikaryotic hyphae, formed after sexual mating of two haploid sporidia of opposite mating type. Therefore, mating/filamentation
Idalis Villanueva et al.
Acta biomaterialia, 5(8), 2832-2846 (2009-06-11)
The pericellular matrix (PCM) surrounding chondrocytes is thought to play an important role in transmitting biochemical and biomechanical signals to the cells, which regulates many cellular functions including tissue homeostasis. To better understand chondrocytes interactions with their PCM, three-dimensional poly(ethylene
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service