Skip to Content
Merck
All Photos(1)

Key Documents

235253

Sigma-Aldrich

(Diethylamino)sulfur trifluoride

95%

Synonym(s):

DAST, Diethylaminosulfur trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NSF3
CAS Number:
Molecular Weight:
161.19
Beilstein:
1849066
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

bp

30-32 °C/3 mmHg (lit.)

density

1.22 g/mL at 25 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CCN(CC)S(F)(F)F

InChI

1S/C4H10F3NS/c1-3-8(4-2)9(5,6)7/h3-4H2,1-2H3

InChI key

CSJLBAMHHLJAAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylamino sulfur trifluoride (DAST) is a fluorinating reagent used in the synthesis of fluorinated compounds and ring-opening reactions.

Application

  • Fluorinating agent: reaction with alcohols and carbonyl compounds, Review
  • Review on nucleophilic fluorination.
  • Catalyst for Friedel-Crafts allylation using tertiary cyclopropyl silyl ethers and the rearrangement of homoallylic alcohols to unsaturated aldehydes.
  • Early introduction of a fluoromethyl group stabilizes the epoxide during further manipulations in the synthesis of 26-fluoro-epothilone.
  • Fluorinating agent for a variety of compounds, including thioethers, alkenols, and cyanohydrins.
  • Reagent for gem difluorination of ketopipecolinic acids.

replaced by

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Self-react. D - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S De Jonghe et al.
Bioorganic & medicinal chemistry letters, 9(21), 3159-3164 (1999-11-24)
Short-chain 3-fluoro-(dihydro)ceramide analogues are synthesized from L-serine using diethylaminosulfur trifluoride (DAST) as fluorinating agent. The apoptogenic activity of these compounds was measured in three different cell lines and compared with their hydroxylated counterparts.
Stereoselective introduction of a trifluoromethyl group into an unsaturated system
Tellier, F.; Sauvetre, R.
Tetrahedron Letters, 32, 5963-5963 (1991)
Nucleic acid related compounds. 79. Efficient conversions of thioethers to α-fluoro thioethers with DAST or DAST/antimony(III) chloride
The Journal of Organic Chemistry, 58, 3800-3800 (1993)
M Kirihara et al.
Chemical & pharmaceutical bulletin, 48(2), 220-222 (2000-03-08)
Diethylaminosulfur trifluoride reacted with cyclic ketoximes bearing substituent(s) that can stabilize a carbocation to cause fluorinative fragmentation, affording fluorinated carbonitrile. Ketoximes lacking such substituents afforded complex mixtures. However, the introduction of a sulfur functionality, which can stabilize a carbocation and
Fluorination with diethylaminosulfur trifluoride and related aminofluorosulfuranes
M. Hudlicky
Org. React., 35, 513-513 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service