900180
Monodispersed nanodiamond particles
5 nm avg. part. size (DLS), 10 mg/mL in H2O, carboxylated
Synonym(s):
Detonation Nanodiamond, Nanodiamond
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352302
NACRES:
NA.23
Recommended Products
description
-45.1 mV ±20%
Quality Level
form
dispersion
nanoparticles
concentration
10 mg/mL in H2O
avg. part. size
5 nm (DLS)
Looking for similar products? Visit Product Comparison Guide
Application
Detonation nanodiamonds find applications in polymer composites for providing improved mechanical properties, UV protection, and improved resistance to radiation. They also find biological applications extending from drug delivery to positron emission tomography and other imaging techniques .
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cancer nanomedicine: from drug delivery to imaging.
Chow E, et al.
Science Translational Medicine, 5 (2013)
Nanodiamond Particles: Properties and Perspectives for Bioapplications.
Schrand AM, et al.
Critical Reviews in Solid State and Materials Sciences, 34, 18-18 (2009)
The properties and applications of nanodiamonds.
Mochalin VN,et al.
Nature Nanotechnology, 7, 11-11 (2012)
Functionality is Key: Recent Progress in the Surface Modification of Nanodiamond.
Krueger A and Lang D
Advances in Functional Materials, 22, 890-890 (2012)
Nanodiamond-mediated drug delivery and imaging: challenges and opportunities.
Vaijayanthimala V, et al.
Expert Opinion on Drug Delivery, 12, 1-1 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service