900173
Fluorescent nanodiamond
Nitrogen vacancy >60 NVN/particle, 70 nm avg. part. size (DLS), 1 mg/mL in deionized water
Synonym(s):
Nanodiamond
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352302
NACRES:
NA.23
Recommended Products
description
+56.7 mV ±20%
Quality Level
form
dispersion
nanoparticles
composition
Nitrogen vacancy, >60 NVN/particle
concentration
1 mg/mL in deionized water
avg. part. size
70 nm (DLS)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Green fluorescent nanodiamond.
Application
Nanodiamonds containing N-V-N color centers exhibit green fluorescence. These do not photobleach or photoblink and find applications in fluorescent bioimaging, cathode luminescent imaging, stimulated emission depletion (STED) imaging etc. In combination with NDs emitting in red fluorescence (N-V), it provides opportunities for multicolor imaging.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Romana Schirhagl et al.
Annual review of physical chemistry, 65, 83-105 (2013-11-28)
Crystal defects in diamond have emerged as unique objects for a variety of applications, both because they are very stable and because they have interesting optical properties. Embedded in nanocrystals, they can serve, for example, as robust single-photon sources or
Yuen Yung Hui et al.
Optics express, 18(6), 5896-5905 (2010-04-15)
Dynamics of fluorescent diamond nanoparticles in HeLa cells has been studied with two-photon fluorescence correlation spectroscopy (FCS). Fluorescent nanodiamond (FND) is an excellent fluorescent probe for bioimaging application, but they are often trapped in endosomes after cellular uptake. The entrapment
Functionality is Key: Recent Progress in the Surface Modification of Nanodiamond.
Krueger A and Lang D
Advances in Functional Materials, 22, 890-890 (2012)
Nanodiamond-mediated drug delivery and imaging: challenges and opportunities.
Vaijayanthimala V, et al.
Expert Opinion on Drug Delivery, 12, 1-1 (2014)
Highly Fluorescent Nanodiamonds Protein-Functionalized for Cell Labeling and Targeting.
Chang B, et al.
Advances in Functional Materials, 23, 5737-5737 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service