900171
Fluorescent nanodiamond
Nitrogen vacancy ~3 ppm, 100 nm avg. part. size (APS), aminated powder
Synonym(s):
Diamond Nanopowder, Nanodiamond
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Zeta potential +42.6 mV ±10%
Quality Level
form
nanoparticles
powder
composition
Nitrogen vacancy, ~3 ppm
avg. part. size
100 nm (APS)
Looking for similar products? Visit Product Comparison Guide
Application
Nanodiamonds containing N-V-N color centers exhibit green fluorescence. These do not photobleach or photoblink and find applications in fluorescent bioimaging, cathodoluminescent imaging, stimulated emission depletion (STED) imaging etc. In combination with NDs emitting in red fluorescence (N-V), it provides opportunities for multicolor imaging.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Annual review of physical chemistry, 65, 83-105 (2013-11-28)
Crystal defects in diamond have emerged as unique objects for a variety of applications, both because they are very stable and because they have interesting optical properties. Embedded in nanocrystals, they can serve, for example, as robust single-photon sources or
Optics express, 18(6), 5896-5905 (2010-04-15)
Dynamics of fluorescent diamond nanoparticles in HeLa cells has been studied with two-photon fluorescence correlation spectroscopy (FCS). Fluorescent nanodiamond (FND) is an excellent fluorescent probe for bioimaging application, but they are often trapped in endosomes after cellular uptake. The entrapment
Functionality is Key: Recent Progress in the Surface Modification of Nanodiamond.
Advances in Functional Materials, 22, 890-890 (2012)
Nanodiamond-mediated drug delivery and imaging: challenges and opportunities.
Expert Opinion on Drug Delivery, 12, 1-1 (2014)
Nanodiamond bioconjugate probes and their collection by electrophoresis.
Diamond and Related Materials, 17, 1858-1858 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service