Skip to Content
Merck
All Photos(1)

Documents

900174

Sigma-Aldrich

Fluorescent nanodiamond

Nitrogen vacancy 3 ppm NV/particle, 100 nm avg. part. size (DLS), 1 mg/mL in deionized water, hydroxylated

Synonym(s):

Nanodiamond

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

Quality Level

form

nanoparticles

composition

Nitrogen vacancy, 3 ppm NV/particle

concentration

1 mg/mL in deionized water

avg. part. size

100 nm (DLS)

Looking for similar products? Visit Product Comparison Guide

Application

Nanodiamonds containing nitrogen-vacancy (NV) centers can be prepared using irradiation with high energy electrons Nanodiamonds with low numbers of NV centers (ideally, single defect centers) are suitable for quantum signaling, optics, and computing applications because their fluorescence allows for observation of single quantum systems.

Storage Class Code

10 - Combustible liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High yield fabrication of fluorescent nanodiamonds.
Boudou JP, et al.
Nanotechnology, 20 (2009)
Single defect centres in diamond: A review.
Jelezko F, et al.
Physica Status Solidi, 203(13), 3207-3225 (2006)
Scanning Confocal Optical Microscopy and Magnetic Resonance on Single Defect Centers.
Gruber A, et al.
Science, 276 (1997)
In Commercial quantities of ultrasmall fluorescent nanodiamonds containing color centers.
Shenderova O, et al.
Advances in Photonics of Quantum Computing, Memory, and Communication X, San Francisco, SPIE: San Francisco (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service