Skip to Content
Merck
All Photos(2)

Documents

900152

Sigma-Aldrich

2-[[(2-Carboxyethyl)sulfanylthiocarbonyl]-sulfanyl]propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O4S3
CAS Number:
Molecular Weight:
254.35
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.23

Assay

95% (GC)

Quality Level

form

powder

mp

120-125 °C

General description

  • A stable, trithiocarbonate chain transfer agent (CTA), also referred to as a RAFT agent, which can control chain growth in free radical polymerization producing polymers with well-defined molecular weights and low polydispersities.
  • 2-(2-Carboxyethylsulfanylthiocarbonylsulfanyl)propionic acid may be used to control homo- and co-polymerization of acrylic acid, acrylates, acrylamides, styrenes, methacrylates, and methacrylamides. It is soluble in aqueous and organic media (amphiphilic).

Caution

Storage conditions: Room temperature, protect from light.

Legal Information

Sold for research purposes only: See sigma-aldrich.com/raftlicense. Patents: WO98/01478, WO99/311444.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Evaluation of New Dicarboxylic Acid Functional Trithiocarbonates: RAFT Synthesis of Telechelic Poly(n-butyl acrylate)s.
Wang R, et al.
Macromolecules, 38, 9518-9525 (2005)
Synthesis of Dual-Functional Poly(6-azidohexylmethacrylate) Brushes by a RAFT Agent Carrying Carboxylic Acid End Groups.
Cimen D, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 53, 1696-1706 (2015)
A Continuous Flow Process for the Radical Induced End Group Removal of RAFT Polymers.
Hornung H, et al.
Macromolecular Reaction Engineering, 6, 246-251 (2012)
Substituent Effects on Trithiocarbonates-Mediated Polymerization of Styrene.
Journal of Applied Polymer Science, 111, 2011-2017 (2009)
Thermo-responsive shell cross-linked PMMA-b-P(NIPAAm-co-NAS) micelles for drug delivery.
Chang C, et al.
International Journal of Pharmaceutics, 420(2), 333-340 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service