Skip to Content
Merck
All Photos(2)

Key Documents

415324

Sigma-Aldrich

N-Isopropylacrylamide

97%

Synonym(s):

NIPAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCONHCH(CH3)2
CAS Number:
Molecular Weight:
113.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

bp

89-92 °C/2 mmHg (lit.)

mp

60-63 °C (lit.)

SMILES string

CC(C)NC(=O)C=C

InChI

1S/C6H11NO/c1-4-6(8)7-5(2)3/h4-5H,1H2,2-3H3,(H,7,8)

InChI key

QNILTEGFHQSKFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Isopropylacrylamide (NIPAM) is a biocompatible monomeric unit that can be used in the formation of stimuli responsive polymers due to its temperature sensitive properties, which include temperature-based volumetric and phase changes. These properties change when the temperature of the solution reaches a lower critical solution temperature (LCST).

Application

Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.
NIPAM can be used to prepare poly(NIPAM) based thermosetting polymers, which can be used for a variety of applications such as tissue engineering, cell culture, biomedical coating, drug delivery, and muscle regeneration.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterisation of biomedical coatings
Coatings for Biomedical Applications, 32(5), 176-220 (2012)
Temperature-responsive polymers for cell culture and tissue engineering applications
Switchable and Responsive Surfaces and Materials for Biomedical Applications, 32(5), 203-233 (2015)
Bio-Instructive Scaffolds for Muscle Regeneration: NonCrosslinked Polymers
Bio-Instructive Scaffolds for Musculoskeletal Tissue Engineering and Regenerative Medicine, 34(9), 161-186 (2017)
Firdaus Yhaya et al.
Macromolecular rapid communications, 33(21), 1868-1874 (2012-08-24)
A block copolymer based on poly(N-isopropyl acrylamide) (PNIPAAm) and a block with a statistical distribution of poly(2-hydroxyethyl acrylate) (PHEA) and repeating unit with carrying β-cyclodextrin was prepared via reversible addition-fragmentation chain transfer (RAFT) polymerization and click reaction. Addition of poly(2-hydroxyethyl
Saunders, B.R.
Surfactant Science Series, 115, 419-419 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service