Skip to Content
Merck
All Photos(1)

Key Documents

751138

Sigma-Aldrich

1-(Methoxycarbonyl)ethyl benzodithioate

≥97%

Synonym(s):

2-[(Phenylthioxomethyl)thio]propanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O2S2
CAS Number:
Molecular Weight:
240.34
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

≥97%

form

liquid

refractive index

n20/D 1.6288

bp

275.2-284.4 °C

density

1.1777 g/mL at 25 °C

storage temp.

2-8°C

InChI

1S/C11H12O2S2/c1-8(10(12)13-2)15-11(14)9-6-4-3-5-7-9/h3-8H,1-2H3

InChI key

WPFPXUQEUXJYHU-UHFFFAOYSA-N

Application

RAFT agent for controlled radical polymerization; well-suited for polymerization of methacrylates, methacrylamides, and to a lesser extent styrenes, acrylates, and acrylamides. Chain Transfer Agent (CTA)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service