Skip to Content
Merck
All Photos(3)

Documents

723274

Sigma-Aldrich

4-Cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentanoic acid

97% (HPLC)

Synonym(s):

4-Cyano-4-(((dodecylthio)carbonothioyl)thio)pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H33NO2S3
CAS Number:
Molecular Weight:
403.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97% (HPLC)

form

solid

mp

64-68 °C

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCSC(=S)SC(C)(CCC(O)=O)C#N

InChI

1S/C19H33NO2S3/c1-3-4-5-6-7-8-9-10-11-12-15-24-18(23)25-19(2,16-20)14-13-17(21)22/h3-15H2,1-2H3,(H,21,22)

InChI key

RNTXYZIABJIFKQ-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

De-Xiang Zhang et al.
Scientific reports, 9(1), 1367-1367 (2019-02-06)
A new and facile approach to selectively functionalize the internal and external surfaces of porous silicon (pSi) for drug delivery applications is reported. To provide a surface that is suitable for sustained drug release of the hydrophobic cancer chemotherapy drug
Tong Yang et al.
Biomaterials science, 8(20), 5698-5714 (2020-09-16)
The combination therapy of cisplatin (CDDP) and metformin (MET) is a clinical strategy to enhance therapeutic outcomes in lung cancer. However, the efficacy of this combination is limited due to the asynchronous pharmacokinetic behavior of CDDP and MET, used as
Yiyuan Guo et al.
Expert opinion on drug delivery, 17(3), 407-421 (2020-02-06)
Background: Natamycin is the only topical ophthalmic antifungal drug approved by the Food and Drug Administration (FDA) of the United States, but has unsatisfactory factors such as high dosing frequency.Methods: We report the synthesis and preparation of self-assembled poly(ethylene glycol)-block-poly(glycidyl
Katharina Hendrich et al.
Polymers, 12(6) (2020-05-30)
Linear and four-arm star polystyrene samples prepared by RAFT polymerization were grafted to gold surfaces directly via their thiocarbonylthio-end groups. Nanoscale polymer patterns were subsequently formed via constrained dewetting. The patterned polymer films then served as a template for the
RAFT Agent Design and Synthesis
Keddie, D. J.; et al.
Macromolecules, 45, 5321-5342 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service