Skip to Content
Merck
All Photos(4)

Key Documents

805831

Sigma-Aldrich

Dimethylammonium iodide

greener alternative

Synonym(s):

Dimethylamine hydroiodide, Greatcell Solar®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H8IN
CAS Number:
Molecular Weight:
173.00
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

153.85 °C

greener alternative category

SMILES string

CNC.I

InChI

1S/C2H7N.HI/c1-3-2;/h3H,1-2H3;1H

InChI key

JMXLWMIFDJCGBV-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Dimethylammonium iodide (DMAI) is used as an additive for the fabrication of perovskite-based solar cells. It improves the crystal phases and morphologies of the perovskite films, which affect the power conversion efficiency (PCE) of the optoelectronic devices.
The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.Greatcell Solar® is a registered trademark of Greatcell Solar Materials Pty Ltd
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of dimethylammonium iodide in CsPbI3 perovskite fabrication: additive or dopant?
Wang Y, et al.
Angewandte Chemie (International Edition in English), 58(46), 16691-16696 (2019)
An engineered thermal-shift screen reveals specific lipid preferences of eukaryotic and prokaryotic membrane proteins
Nji E, et al.
Nature Communications, 9(1), 1-12 (2018)
Wei Zhang et al.
Nano letters, 15(3), 1698-1702 (2015-02-05)
The performance of perovskite solar cells has been progressing over the past few years and efficiency is likely to continue to increase. However, a negative aspect for the integration of perovskite solar cells in the built environment is that the
Nam Joong Jeon et al.
Nature, 517(7535), 476-480 (2015-01-07)
Of the many materials and methodologies aimed at producing low-cost, efficient photovoltaic cells, inorganic-organic lead halide perovskite materials appear particularly promising for next-generation solar devices owing to their high power conversion efficiency. The highest efficiencies reported for perovskite solar cells
Zhi-Kuang Tan et al.
Nature nanotechnology, 9(9), 687-692 (2014-08-05)
Solid-state light-emitting devices based on direct-bandgap semiconductors have, over the past two decades, been utilized as energy-efficient sources of lighting. However, fabrication of these devices typically relies on expensive high-temperature and high-vacuum processes, rendering them uneconomical for use in large-area

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service