Skip to Content
Merck
All Photos(3)

Documents

805874

Sigma-Aldrich

n-Butylammonium iodide

greener alternative

Synonym(s):

1-Butanaminium iodide, Butylamine hydroiodide, Butylammonium iodide, Greatcell Solar®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12IN
CAS Number:
Molecular Weight:
201.05
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

173 °C (exp.)

greener alternative category

SMILES string

CCCCN.I

InChI

1S/C4H11N.HI/c1-2-3-4-5;/h2-5H2,1H3;1H

InChI key

CALQKRVFTWDYDG-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

n-Butylammonium iodide (BAI) can be used as an additive that facilitates an improvement in the efficiency and stability of perovskite solar cells (PSCs). It can also be used as an organic ligand in the formation of organic-inorganic perovskites for light-emitting diodes (LEDs).
The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced thermal stability in perovskite solar cells by assembling 2D/3D stacking structures
Lin Y, et al.
The Journal of Physical Chemistry Letters, 9(3), 654-658 (2018)
Unveiling the guest effect of N-butylammonium iodide towards efficient and stable 2D-3D perovskite solar cells through sequential deposition process
Wang Y, et al.
Chemical Engineering Journal, 6(2), 123589-123589 (2019)
Structural changes and band gap tunability with incorporation of n-butylammonium iodide in perovskite thin film
Rebecca LWX, et al.
Heliyon, 6(2), e03364-e03364 (2020)
Tunable light-emitting diodes utilizing quantum-confined layered perovskite emitters
Congreve DN, et al.
ACS photonics, 4(3), 476-481 (2017)
Olivia F Williams et al.
The journal of physical chemistry. A, 123(51), 11012-11021 (2019-11-16)
Two-dimensional (2D) hybrid perovskites are generating broad scientific interest because of their potential for use in photovoltaics and microcavity lasers. It has recently been demonstrated that mixtures of quantum wells with different thicknesses can be assembled in films with heterogeneous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service