Skip to Content
Merck
All Photos(4)

Key Documents

806048

Sigma-Aldrich

Formamidinium iodide

greener alternative

Synonym(s):

Greatcell Solar®, Iminomethylamine hydriodide, Methanimidamide iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH5IN2
Molecular Weight:
171.97
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.23

description

Elemental Analysis: C ~7.0%
Elemental Analysis: N ~16.3%

Quality Level

Assay

≥98% (H-NMR)

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

335 °C

greener alternative category

SMILES string

[NH2+]=C([H])N.[I-]

InChI

1S/CH4N2.HI/c2-1-3;/h1H,(H3,2,3);1H

InChI key

QHJPGANWSLEMTI-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Formamidinium iodide (FAI) is an organic halide, which can be used as a precursor solution in the fabrication of perovskite-based heterojunction solar cells.
Formamidinium iodide (FAI) serves as a critical precursor material in the fabrication of perovskite solar cells. FAI is used in material engineering studies to investigate the impact of formamidinium incorporation on perovskite film properties and device performance.
The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar® is a registered trademark of Greatcell Solar Materials Pty Ltd
Greatcell Solar is a registered trademark of Greatcell Solar

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dazheng Chen et al.
Nanomaterials (Basel, Switzerland), 9(7) (2019-07-03)
Indium thin oxide (ITO)-free planar perovskite solar cells (PSCs) were fabricated at a low temperature (150 °C) in this work based on the transparent electrode of photolithography processed nickel/gold (Ni/Au) mesh and the high conductivity polymer, PH1000. Ultrathin Au was
High-performance photovoltaic perovskite layers fabricated through intramolecular exchange
Yang WS, et al.
Science, 348(6240), 1234-1237 (2015)
Ihteaz M Hossain et al.
Optics express, 28(6), 8878-8897 (2020-04-01)
The rise in the power conversion efficiency (PCE) of perovskite solar cells has triggered enormous interest in perovskite-based tandem photovoltaics. One key challenge is to achieve high transmission of low energy photons into the bottom cell. Here, nanostructured front electrodes
Long Ji et al.
Nanoscale research letters, 12(1), 367-367 (2017-05-26)
Lead-free solution-processed solid-state photovoltaic devices based on formamidinium tin triiodide (FASnI
Efficient inverted planar formamidinium lead iodide perovskite solar cells via a post improved perovskite layer
Zhang Y, et al.
Royal Society of Chemistry Advances, 6(83), 79952-79957 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service