735140
Fluorine doped tin oxide coated glass slide
L × W × thickness 50 mm × 50 mm × 2.2 mm, surface resistivity ~7 Ω/sq
Synonym(s):
TEC 7, FTO glass
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Haze: 5%
composition
SnO2/F
packaging
set of 5
surface resistivity
~7 Ω/sq
L × W × thickness
50 mm × 50 mm × 2.2 mm
transmittance
80-82% (visible)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The coating thickness of Fluorine doped tin oxide on glass slide is ∼ 550 nm.The glass has a refractive index of ∼ 1.52 and the SnO2:F coating is ∼ 1.85.
Application
- Procyanidin B2 is used as a reference standard: It is used for assessing the proanthocyanidin content in cranberries using 4-(Dimethylamino)cinnamaldehyde Assay. This standard helps in quantifying and characterizing the proanthocyanidin content in cranberries by providing a known benchmark for comparison. (Feliciano et al., 2012).
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nanomaterials (Basel, Switzerland), 10(2) (2020-02-09)
Microtubules are hollow cylindrical polymers composed of the highly negatively-charged (~23e), high dipole moment (1750 D) protein α, β- tubulin. While the roles of microtubules in chromosomal segregation, macromolecular transport, and cell migration are relatively well-understood, studies on the electrical
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service