773506
Ethyl 2-(phenylcarbonothioylthio)-2-phenylacetate
98%
Synonym(s):
α-[(Phenylthioxomethyl)thio]benzeneacetic acid ethyl ester, dithiobenzoate RAFT, Benzodithioate phenyl acetic acid ethyl ester
About This Item
Recommended Products
Assay
98%
form
solid
mp
48-53 °C
storage temp.
2-8°C
SMILES string
CCOC(=O)C(SC(=S)c1ccccc1)c2ccccc2
InChI
1S/C17H16O2S2/c1-2-19-16(18)15(13-9-5-3-6-10-13)21-17(20)14-11-7-4-8-12-14/h3-12,15H,2H2,1H3
InChI key
SQKNCNYHCGWDEX-UHFFFAOYSA-N
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service