Skip to Content
Merck
All Photos(1)

Documents

723002

Sigma-Aldrich

Cyanomethyl methyl(phenyl)carbamodithioate

98% (HPLC)

Synonym(s):

Cyanomethyl N-methyl-N-phenyl dithiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2S2
CAS Number:
Molecular Weight:
222.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98% (HPLC)

form

powder

mol wt

average Mn 8,000

mp

88-92 °C

storage temp.

2-8°C

SMILES string

CN(C(=S)SCC#N)c1ccccc1

InChI

1S/C10H10N2S2/c1-12(10(13)14-8-7-11)9-5-3-2-4-6-9/h2-6H,8H2,1H3

InChI key

FYACMHCOSCVNHO-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization; especially suited for the polymerization of vinyl ester and vinyl amide monomers. Chain Transfer Agent (CTA)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RAFT Agent Design and Synthesis
Keddie, D. J.; et al.
Macromolecules, 45, 5321-5342 (2012)
Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service