Skip to Content
Merck
All Photos(1)

Documents

765430

Sigma-Aldrich

Gold nanoparticles

5 nm diameter, carboxylic acid functionalized, PEG 3000 coated, OD 50, dispersion in H2O

Synonym(s):

Gold nanoparticles COOH functionalized, Au NP COOH, Gold Colloid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

material

PEG 3000

form

dispersion in H2O
nanoparticles

packaging

poly bottle of 1 mL

OD

50

diameter

5 nm

pH

6.0-8.0 (25 °C)

solubility

water: miscible

density

1.00 g/cm3

λmax

515 nm

functional group

carboxylic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Carboxylic acid functionalized GNPs forms conjugates with various protein molecules due to the electrostatic attraction between negatively charged carboxylate groups and positively charged amino groups of protein.

Application

Carboxylic acid functionalized, PEG 3000 coated, OD 50, and H2O dispersed gold nanoparticles (5 nm) can be potentially used in the development of biosensors, cancer-cell imaging, drug delivery, and photothermal therapy.
AuNPs may be used to prepare SARS-CoV-2 conjugate for COVID-19 detection using lateral flow immunoassay technique. It may also be used in the immobilization of antibodies to improve the sensitivity of optical fiber sensors for the screening of COVID-19. It can also be deposited on the titanium electrode for the formation of a miniaturized device for the electrochemical detection of DNA hybridization.
Carboxylic acid group binds with various proteins and is used in targeted drug delivery and gene therapy.
Functionalized Particles have been extensively used to study cellular uptake of nanoparticles and targeted drug delivery.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nóra Adányi et al.
Food chemistry, 267, 10-14 (2018-06-24)
Mycotoxins, present in a wide range of food and feed commodities, are toxic secondary metabolites produced by a number of different fungi. Certain mycotoxins do not readily degrade at high temperatures, therefore are resistant to food processing, and consequently are
Development and clinical application of a rapid IgM-IgG combined antibody test for SARS-CoV-2 infection diagnosis
Li Z, et al.
Journal of Medical Virology, 6, 2071-2071 (2020)
Size-dependent in vivo toxicity of PEG-coated gold nanoparticles
Zhang X, et al.
International journal of nanomedicine, 6, 2071-2071 (2011)
Nishima Wangoo et al.
Journal of colloid and interface science, 323(2), 247-254 (2008-05-20)
We report a novel strategy for the synthesis of aqueous stable, carboxylated gold nanoparticles (GNPs) by using glutamic acid as the reducing agent. The ratio of chloroaurate ions, AuCl(-)(4) to glutamic acid was optimized in the reaction medium to obtain
Recent advances in nanoparticle-based lateral flow immunoassay as a point-of-care diagnostic tool for infectious agents and diseases
Banerjee R and Jaiswal A
Analyst, 143(9), 1970-1996 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service