Skip to Content
Merck
All Photos(1)

Key Documents

741965

Sigma-Aldrich

Gold nanoparticles

20 nm diameter, OD 1, stabilized suspension in citrate buffer

Synonym(s):

Au NP, Gold Colloid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanoparticles
suspension

contains

Proprietary Surfactant as stabilizer

concentration

~6.54E+11 particles/mL

OD

1

diameter

20 nm

λmax

518-522 nm

PDI

<0.2

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanoparticles (AuNPs) are versatile materials widely used in biomedical sciences and electrochemical sensor based applications. AuNPs are tunable nano-materials with excellent thermo-mechanical properties, high surface area, and low toxicity.

Application

Citrate stabilized AuNPs can be used for a variety of applications such as:
  • neutrophil labeling for monitoring the effect of different drugs
  • immobilization of proteins for biosensor applications
  • nano-carrier for drug delivery system for the release of hydrophobic drugs.

Ideal for adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications. Note: This product is not ideal for protein adsorption. For gold nanoparticles suitable for adsorption of proteins and other ligands for conjugate development, please see our gold nanoparticles stabilized in PBS.

This material is highly monodisperse (<12% variability in size and shape), and provides significantly improved surface reactivity. Applications include Surface Enhanced Raman Lables, Sensing/Detection, Biological Targeting, Plasmonics and Electronics.

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced thermomechanical properties of a nematic liquid crystal elastomer doped with gold nanoparticles
Montazami R, et al.
Sensors and actuators A, Physical, 178(11), 175-178 (2012)
Gold nanoparticles in delivery applications
Ghosh P, et al.
Advanced Drug Delivery Reviews, 60(11), 1307-1315 (2008)
Toxicity and cellular uptake of gold nanoparticles: what we have learned so far?
Alkilany AM and Murphy CJ
Journal of Nanoparticle Research, 12(7), 2313-2333 (2010)
Evaluation of novel Fmoc-tripeptide based hydrogels as immobilization supports for electrochemical biosensors
Fusco G, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 137(24), 105-110 (2018)
Gold-Organic-Inorganic High-Surface-Area Materials as Precursors of Highly Active Catalysts
Budroni G and Corma A
Angewandte Chemie (International ed. in English), 45(20), 3328-3331 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service