765341
Gold nanoparticles
20 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
Synonym(s):
Gold nanoparticles NH2 functionalized, Au NP NH2, Gold Colloid
About This Item
Recommended Products
material
PEG 5000
Quality Level
form
dispersion in H2O
nanoparticles
packaging
poly bottle of 1 mL
OD
50
diameter
20 nm
pH
6.0-8.0 (25 °C)
solubility
water: miscible
density
1.00 g/cm3
λmax
520 nm
functional group
amine
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Since they adhere to cell membranes, AuNPs are used in cellular and intracellular targeting in targeted drug delivery applications and may also be used in biodistribution studies
- AuNPs may also be used in photothermal therapy and radiotherapy.
- Proposed applications of gold nanoparticles are as:drug delivery agents for cancer therapy, coatings, plastics, nanowire, nanofiber, textiles, catalyst applications.
- They may also be used in carrying siRNA which acts against human prostate carcinoma cells by inhibiting a specific cancer gene.[5][4]
Features and Benefits
- The negatively charged siRNA-PEG complex, attached to positively charged GNPs, is easily cleavable in reductive cytosolic environment thus enabling the release of siRNA into cytosol.[5][4]
- The PEG coating decreases the cytotoxicity and increases effciciency of GNPs. PEG increases the stability of the nanoparticles and prevents agglomeration.[1]
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service