Skip to Content
Merck
All Photos(2)

Key Documents

750530

Sigma-Aldrich

Carbon nanotube, single-walled

98% (Metallic), avg. no. of layers, 1

Synonym(s):

IsoNanotubes-M, SWCNT, SWNT, Single wall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

Assay

2% (Semiconducting)
98% (Metallic)

form

solid

feature

avg. no. of layers 1

manufacturer/tradename

NanoIntegris, Inc.

L

0.3-5 μm

diameter

1.2-1.7 nm

mp

3652-3697 °C (lit.)

density

1.7-1.9 g/cm3 at 25 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

IsoNanotubes-M are produced by arc discharge or plasma torch. The diameter of the semiconducting single walled nanotubes range in 1.2-1.7 nm, whereas the length varies from 100nm to 4 μm. The degree of purity of the SWNTs can be determined from the absorbance spectrum. The optical absorbance of metallic SWNTs occurs at M11 peaks (550-800nm).

Application

These carbon nanotubes are produced via the Arc Discharge process and are purified via density gradient centrifugation.

Research applications include:
  • Electronics
  • Sensors
  • Composites
  • Energy Storage
  • Study of Life Science systems

Preparation Note

Electric Arc Discharge Method

Legal Information

Product of NanoIntegris, Inc
IsoNanotubes-M is a trademark of NanoIntegris, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuan Wang et al.
ACS nano, 4(12), 7123-7132 (2010-11-11)
Macroelectronic integrated circuits are widely used in applications such as flat panel display and transparent electronics, as well as flexible and stretchable electronics. However, the challenge is to find the channel material that can simultaneously offer low temperature processing, high
Chad D Vecitis et al.
ACS nano, 4(9), 5471-5479 (2010-09-04)
Single-walled carbon nanotubes (SWNTs) have been previously observed to be strong antimicrobial agents, and SWNT coatings can significantly reduce biofilm formation. However, the SWNT antimicrobial mechanism is not fully understood. Previous studies on SWNT cytotoxicity have concluded that membrane stress
Amin Salehi-Khojin et al.
ACS nano, 5(1), 153-158 (2010-12-29)
There has been recent controversy whether the response seen in carbon nanotube (CNT) chemiresistors is associated with a change in the resistance of the individual nanotubes or changes in the resistance of the junctions. In this study, we carry out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service