698849
Carbon nanotube, multi-walled
>98% carbon basis, O.D. × L 6-13 nm × 2.5-20 μm
Synonym(s):
MWCNT, MWNT, Multiwall carbon nanotube
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
Assay
>98% carbon basis
form
powder
composition
carbon, >98% (total metals impurities)
O.D. × L
6-13 nm × 2.5-20 μm
size
10 μm , average length, TEM
12 nm , average diameter, HRTEM
surface area
~220 m2/g , SSA
mp
3652-3697 °C (lit.)
density
~2.1 g/mL at 25 °C (lit.)
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Multi walled carbon nanotubes (MWNTs, CNTs) were prepared by chemical vapor deposition (CVD). In chemical vapor deposition (CVD), a volatile precursor undergoes thermal decomposition at elevated temperatures to form a solid deposit on a substrate. The diameter of the tube ranges between 6-13nm and the average length is 10μm. The multiwalled nanotubes are stable in inert atmosphere upto a temperature of 3697oC. CVD production method is followed by HCl demineralization.
Application
A hybrid nanostructure system of MWNTs/titania was fabricated to be used as photodiodes in dye sensitized solar cells. MWNTs were investigated as electrodes for electrochemical biosensors. Use of MWNTs increased the efficacy, sensitivity and mechanical strength of the sensors.
Carbon nanotube, multi-walled (MWNT) belongs to the class of carbonaceous materials with excellent physiochemical, thermo-mechanical and electrochemical properties. This material can be used in a variety of sustainable energy applications such as solar cells, photocatalysis, biosensor, gas sensor, supercapacitor and as a filler that acts as a reinforcement to improve the mechanical property of composites.
Physical form
average wall thickness 7-13 graphene layers
Preparation Note
Chemical Vapor Deposition (CVD) Method
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A microfluidic chip platform with electrochemical carbon nanotube electrodes for pre-clinical evaluation of antibiotics nanocapsules.
Hong CC, et al.
Biosensors And Bioelectronics, 26(8), 3620-3626 (2011)
Hybrid carbon nanotubes?TiO2 photoanodes for high efficiency dye-sensitized solar cells.
Dembele KT, et al.
The Journal of Physical Chemistry C, 117(28), 14510-14517 (2013)
Jin Wuk Lee et al.
Aquatic toxicology (Amsterdam, Netherlands), 171, 9-19 (2015-12-31)
Multi-walled carbon nanotubes (MWCNTs) are nanoparticles widely applicable in various industrial fields. However, despite the usefulness of MWCNTs in industry, their oxidative stress-induced toxicity, combined toxicity with metal, and mitogen-activated protein kinase (MAPK) activation have not been widely investigated in
Si Gao et al.
Nature communications, 8(1), 163-163 (2017-08-02)
Knowing the three-dimensional structural information of materials at the nanometer scale is essential to understanding complex material properties. Electron tomography retrieves three-dimensional structural information using a tilt series of two-dimensional images. In this paper, we report an alternative combination of
Effect of the method of dispersion on the properties of carbon nanotube and microfibrillar cellulose composites
Nadera A
Inventia Report, 433 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service