Skip to Content
Merck
All Photos(2)

Documents

201375

Sigma-Aldrich

Brilliant Yellow

Dye content ≥50 %

Synonym(s):

Direct Yellow 4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H18N4Na2O8S2
CAS Number:
Molecular Weight:
624.55
Colour Index Number:
24890
Beilstein:
4122338
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, ≥50%

technique(s)

titration: suitable

visual transition interval

6.5-8.0, orange to red-orange

λmax

497 nm at 0.1 N in NaOH

ε (extinction coefficient)

≥37000 at 260-268 nm
≥37000 at 486-494 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1ccc(cc1)\N=N\c2ccc(\C=C\c3ccc(cc3S([O-])(=O)=O)\N=N\c4ccc(O)cc4)c(c2)S([O-])(=O)=O

InChI

1S/C26H20N4O8S2.2Na/c31-23-11-7-19(8-12-23)27-29-21-5-3-17(25(15-21)39(33,34)35)1-2-18-4-6-22(16-26(18)40(36,37)38)30-28-20-9-13-24(32)14-10-20;;/h1-16,31-32H,(H,33,34,35)(H,36,37,38);;/q;2*+1/p-2/b2-1+,29-27+,30-28+;;

InChI key

YLDIUICHQPKMNH-MAPAHAHLSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Brilliant Yellow is an orange colored powder and it belongs to the group of azo dyes. It is yellow at pH 6.4 and red-orange at pH 8.0. Brilliant Yellow has been widely used as a biosensor, anti-fungal agent, anti-AIDS (acquired immunodeficiency syndrome) agent and is used in assays for enzyme activity.

Application

Brilliant Yellow has been used to determine the transfer of inoculum from male moths to females during mating.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee C West et al.
Organic & biomolecular chemistry, 1(5), 887-894 (2003-08-22)
Complexation of the Brilliant Yellow tetraanion, 3(4-), by two new diazacoronand linked beta-cyclodextrin (beta CD) dimers 4,13-bis(2-(6A-deoxy-beta-cyclodextrin-6A-yl)aminooctylamidomethyl- and 4,13-bis(8-(6A-deoxy-beta-cyclodextrin-6A-yl)aminooctylamidomethyl)-4,13- diaza-1,7,10-trioxacyclopentadecane, 1 and 2, respectively, has been studied in aqueous solution. UV-visible spectrophotometric studies at 298.2 K, pH 10.0 and I
Yuya Egawa et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(8), 1117-1119 (2006-08-10)
Multilayered thin films containing poly(allylamine) (PAA) and brilliant yellow (BY) were prepared on a quartz slide by a layer-by-layer (LBL) deposition technique. The UV-visible spectra of the PAA/BY films were sufficiently changed depending upon the pH value of the solution
Dissemination of entomopathogenic fungi using Buseola fusca male as vector.
Maniania N K
Biological Control, 374-378 (2011)
Alok Mittal et al.
Environmental science and pollution research international, 19(6), 2438-2447 (2012-01-31)
PURPOSE AND AIM: Removal of an anionic azo dye Brilliant Yellow has been carried out from its aqueous solutions by using hen feathers as potential adsorbent. Hen feathers procured from local poultry were cut, washed, and activated. Detailed chemical and
Z Salamon et al.
Journal of photochemistry and photobiology. B, Biology, 3(3), 351-357 (1989-06-01)
In this paper the phenomenon of a photosensitive ion complex of Brilliant Yellow and ferric ions formation in the electrolyte phase and its subsequent deposition within a bilayer lipid membrane (BLM) is described. Deposition of light sensitive complex into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service