Skip to Content
Merck
All Photos(3)

Key Documents

901634

Sigma-Aldrich

Carbon nanotube, single-walled

Low metal content, avg. no. of layers, 1

Synonym(s):

CNT, Tuball, SWCNT, SWNT, Single wall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

Number of CNT layers: Single layer by TEM

Assay

≥93% (carbon as SWNT by TEM, TGA)
>99% (carbon content, EDX, TGA)

form

powder

feature

avg. no. of layers 1

L

>5 μm , By AFM

diam.

1.2-2.0 nm , By Optical absorption

impurities

<1% Metal impurities (TGA, By EDX)

color

black

mp

3652-3697 °C (lit.)

density

1.7-1.9 g/cm3 at 25 °C (lit.)

SMILES string

[C]

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Low-iron, purified SWCNT powder especially for use in energy applications

Application

Carbon nanotube, single-walled (SWNT) can be used in a variety of applications such as chemical sensing, field-effect transistors (FETs), thin-film transistors (TFTs), hydrogen storage, catalysis and DNA detection.
Carbon nanotube, single-walled (low metal content) can be used for a variety of applications such as heterojunction solar cells, stretchable electronics, photodetector, p-type thermoelectric material, and hydrogen storage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Room temperature methane detection using palladium loaded single-walled carbon nanotube sensors
Lu Y, et al.
Chemical Physics Letters, 391(4-6), 344-348 (2004)
Significant enhancement of single-walled carbon nanotube based infrared photodetector using PbS quantum dots
Tang Y, et al.
IEEE Journal of Selected Topics in Quantum Electronics, 24(6), 1-8 (2018)
High performance p-type organic thermoelectric materials based on metalloporphyrin/single-walled carbon nanotube composite films
Zhou Y, et al.
Journal of Power Sources, 423(6), 152-158 (2019)
Aerosol-jet-printed, high-speed, flexible thin-film transistor made using single-walled carbon nanotube solution
Jones CS, et al.
Microelectronic Engineering, 87(3), 434-437 (2010)
Growth of vertically aligned single-walled carbon nanotube films on quartz substrates and their optical anisotropy
Murakami Y, et al.
Chemical Physics Letters, 385(3-4), 298-303 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service