Skip to Content
Merck
All Photos(1)

Documents

735787

Sigma-Aldrich

Barium

beads, 0.5-2.0 mm particle size, 99% trace metals basis

Synonym(s):

Barium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ba
CAS Number:
Molecular Weight:
137.33
EC Number:
MDL number:
UNSPSC Code:
12141505
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99% trace metals basis

form

beads

resistivity

50.0 μΩ-cm, 20°C

particle size

0.5-2.0 mm

bp

1640 °C (lit.)

mp

725 °C (lit.)

density

3.6 g/mL at 25 °C (lit.)

SMILES string

[Ba]

InChI

1S/Ba

InChI key

DSAJWYNOEDNPEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Critical for the synthesis of pure Ba21M2O5HX (M = Ge, SI, Ga, In, Tl) suboxide Zintl phases.3

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Haifeng Zheng et al.
American journal of physiology. Cell physiology, 306(7), C705-C713 (2014-01-31)
Interstitial cells of Cajal (ICC) generate slow waves in gastrointestinal (GI) muscles. Previous studies have suggested that slow wave generation and propagation depends on a voltage-dependent Ca(2+) entry mechanism with the signature of a T-type Ca(2+) conductance. We studied voltage-dependent
Christine Austin et al.
Nature, 498(7453), 216-219 (2013-05-24)
Early-life dietary transitions reflect fundamental aspects of primate evolution and are important determinants of health in contemporary human populations. Weaning is critical to developmental and reproductive rates; early weaning can have detrimental health effects but enables shorter inter-birth intervals, which
André Deutschmann et al.
Molecular pharmacology, 83(2), 501-511 (2012-12-01)
Bisphenol A (BPA), a high volume production chemical compound attracts growing attention as a health-relevant xenobiotic in humans. It can directly bind to hormone receptors, enzymes, and ion channels to become biologically active. In this study we show that BPA
Ahmed Nagy et al.
Dysphagia, 30(4), 445-451 (2015-06-07)
The aim of this study was to determine whether measures of hyoid velocity increase when swallowing liquids of thicker consistency at a constant volume. A gender-balanced sample of 20 healthy young participants (mean age 31.5) each swallowed 3 boluses of
Shavkat Mamatkulov et al.
The Journal of chemical physics, 138(2), 024505-024505 (2013-01-17)
We develop force field parameters for the divalent cations Mg(2+), Ca(2+), Sr(2+), and Ba(2+) for molecular dynamics simulations with the simple point charge-extended (SPC/E) water model. We follow an approach introduced recently for the optimization of monovalent ions, based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service