Skip to Content
Merck
All Photos(2)

Documents

461210

Sigma-Aldrich

Cerium

chips

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ce
CAS Number:
Molecular Weight:
140.12
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

Quality Level

form

chips
chunks

reaction suitability

reagent type: catalyst
core: cerium

resistivity

73 μΩ-cm, 20°C

bp

3443 °C (lit.)

mp

795 °C (lit.)

density

6.67 g/mL at 25 °C (lit.)

SMILES string

[Ce]

InChI

1S/Ce

InChI key

GWXLDORMOJMVQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jan M Tomczak et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(3), 904-907 (2013-01-11)
Many inorganic pigments contain heavy metals hazardous to health and environment. Much attention has been devoted to the quest for nontoxic alternatives based on rare-earth elements. However, the computation of colors from first principles is a challenge to electronic structure
David Ecija et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(17), 6678-6681 (2013-04-12)
The tessellation of the Euclidean plane by regular polygons has been contemplated since ancient times and presents intriguing aspects embracing mathematics, art, and crystallography. Significant efforts were devoted to engineer specific 2D interfacial tessellations at the molecular level, but periodic
Joanna M Ogiegło et al.
The journal of physical chemistry. A, 117(12), 2479-2484 (2013-03-09)
The optical properties of gadolinium gallium aluminum garnet, Gd3(Ga,Al)5O12, doped with Ce(3+) are investigated as a function of the Ga/Al ratio, aimed at an improved understanding of the energy flow and luminescence quenching in these materials. A decrease of both
Ajay Kumar Yagati et al.
Biosensors & bioelectronics, 47, 385-390 (2013-04-24)
In this study, an enzymatic biosensor for amperometric detection of hydrogen peroxide was developed based on the direct electrochemistry of myoglobin (Mb) on a porous cerium dioxide (CeO2) nanostructured film. The developed film accomplished with large surface area was electrodeposited
C J Van Noorden et al.
Journal of microscopy, 171(Pt 1), 3-16 (1993-07-01)
Methods based on the use of cerium to detect the activity of oxidases and phosphatases are rapidly expanding. Both classes of enzymes can be demonstrated with cerium at the electron and light microscopical level. The in situ detection of H2O2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service