Skip to Content
Merck
All Photos(2)

Documents

403326

Sigma-Aldrich

Strontium

granular, 99% trace metals basis

Synonym(s):

Strontium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sr
CAS Number:
Molecular Weight:
87.62
EC Number:
MDL number:
UNSPSC Code:
12141504
PubChem Substance ID:
NACRES:
NA.23

description

under oil

Assay

99% trace metals basis

form

granular

reaction suitability

reagent type: reductant

resistivity

23 μΩ-cm, 20°C

bp

1384 °C (lit.)

mp

757 °C (lit.)

density

2.6 g/mL at 25 °C (lit.)

SMILES string

[Sr]

InChI

1S/Sr

InChI key

CIOAGBVUUVVLOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to make Strontium-potassium-niobate (Sr2KNb5O15, SKN). SKN is of great interest for pyroelectric sensors and various electro-optic devices.2

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2 - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily Y Tsui et al.
Nature chemistry, 5(4), 293-299 (2013-03-21)
Redox-inactive metals are found in biological and heterogeneous water oxidation catalysts, but, at present, their roles in catalysis are not well understood. Here, we report a series of high-oxidation-state tetranuclear-dioxido clusters comprising three manganese centres and a redox-inactive metal (M).
Zuzana Saidak et al.
Pharmacology & therapeutics, 136(2), 216-226 (2012-07-24)
Osteoporosis is an important age-related bone disease characterized by increased bone turnover with insufficient bone formation relative to bone resorption. According to the current understanding of this disorder, anti-resorptive and anabolic drugs have been developed for therapeutic intervention. Another therapeutic
Faisal Hammad Mekky Koua et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(10), 3889-3894 (2013-02-22)
Oxygen-evolving complex of photosystem II (PSII) is a tetra-manganese calcium penta-oxygenic cluster (Mn4CaO5) catalyzing light-induced water oxidation through several intermediate states (S-states) by a mechanism that is not fully understood. To elucidate the roles of Ca(2+) in this cluster and
Wenjie Zhang et al.
Biomaterials, 34(13), 3184-3195 (2013-02-06)
Both surface chemistry and topography have significant influence on good and fast osseointegration of biomedical implants; the main goals in orthopeadic, dental and maxillofacial surgeries. A surface modification strategy encompassing the use of bioactive trace elements together with surface micron/nano-topographical
A Schaafsma et al.
Critical reviews in food science and nutrition, 41(4), 225-249 (2001-06-13)
For early prevention or inhibition of postmenopausal and age-related bone loss, nutritional interventions might be a first choice. For some vitamins and minerals an important role in bone metabolism is known or suggested. Calcium and vitamin D support bone mineral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service