Skip to Content
Merck
All Photos(3)

Documents

266639

Sigma-Aldrich

Cobalt

powder, 2 μm particle size, 99.8% trace metals basis

Synonym(s):

Co

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.8% trace metals basis

form

powder

reaction suitability

core: cobalt
reagent type: catalyst

resistivity

6.24 μΩ-cm, 20°C

particle size

2 μm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cobalt powder has been used in the preparation of cobalt-based alloys and composites as anode materials.
It can be also used:
  • In the preparation of soft magnetic materials by powder metallurgical processes.
  • As a catalyst in the transformation of ethanol to 1-butanol.

Analysis Note

Average particle size determined by Fisher sub-sieve sizer

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1B - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hydrothermal synthesis of 1-butanol from ethanol catalyzed with commercial cobalt powder
Zhang X, et al.
ACS sustainable chemistry & engineering, 1(12), 1493-1497 (2013)
Preparation and properties of Cobalt-based soft magnetic material prepared by novel powder metallurgy.
Srivastava, Yogesh and Srivastava, Sanjay
Journal of magnetism and magnetic materials, 423(1), 267-274 (2017)
Characterization of amorphous and crystalline tin-cobalt anodes.
Fan, Quan and Chupas, Peter J and Whittingham, M Stanley
Electrochemical and Solid-State Letters, 10(12), A274-A278 (2007)
Electrode properties of Mg2Ni alloy ball-milled with cobalt powder.
Chen, J and Bradhurst, DH and Dou, SX and Liu, HK
Electrochimica Acta, 44(2-3), 353-355 (1998)
Cobalt-silica magnetic nanoparticles with functional surfaces.
Vadala ML, et al.
Journal of magnetism and magnetic materials, 293(1), 162-170 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service