735167
Fluorine doped tin oxide coated glass slide
L × W × thickness 300 mm × 300 mm × 2.2 mm, surface resistivity ~7 Ω/sq
Synonym(s):
FTO glass
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Haze: 5%
Quality Level
composition
SnO2/F
surface resistivity
~7 Ω/sq
L × W × thickness
300 mm × 300 mm × 2.2 mm
transmittance
80-82% (visible)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The coating thickness of Fluorine doped tin oxide on glass slide is ∼ 550 nm. The glass has a refractive index of ∼ 1.52 and the SnO2:F coating is ∼ 1.85.
Application
Fluorine doped tin oxide coated glass slide (FTO glass) can be used as a substrate for the fabrication of dye sensitized solar cells (DSSCs) and photovoltaic cells (PVs).
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Malkeshkumar Patel et al.
Beilstein journal of nanotechnology, 9, 2432-2442 (2018-09-27)
Co3O4 has been widely studied as a catalyst when coupled with a photoactive material during hydrogen production using water splitting. Here, we demonstrate a photoactive spinel Co3O4 electrode grown by the Kirkendall diffusion thermal oxidation of Co nanoparticles. The thickness-dependent
Hydrothermal fabrication of hierarchically anatase TiO 2 nanowire arrays on FTO glass for dye-sensitized solar cells
Wu W, et al.
Scientific reports, 3, 1352-1352 (2013)
Facile solution-controlled growth of CuInS2 thin films on FTO and TiO2/FTO glass substrates for photovoltaic application
Peng S, et al.
Journal of alloys and compounds, 481(1-2), 786-791 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service