Skip to Content
Merck
All Photos(2)

Documents

636916

Sigma-Aldrich

Indium tin oxide coated glass slide, rectangular

surface resistivity 15-25 Ω/sq, slide

Synonym(s):

ITO, ITO coated slide, rectangular

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

surface resistivity

15-25 Ω/sq

L × W × thickness

75 mm × 25 mm × 1.1 mm

transmittance

>78%

refractive index

n20/D 1.517 (lit.)

SMILES string

O=[Sn]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium tin oxide (ITO) is widely used as transparent conducting oxides. Its major characteristics are electrical conductivity, optical transparency and the ease to be deposited as films. ITO coated glass has a sheet resistance of 16Ω/cm2.

Application

ITO coated glass slides were used as substrates for barium vanadate films, α- and β-Fe2O3 nanostructures. The product was used in a study of photoelectrical properties of spin coated polyphenylene films. Poly(N-vinylcarbazole) was spin coated onto the substrate to form a light emitting layer towards the fabrication of light emitting diode (LED).

Physical properties

Thickness of ITO coating is 600-1000 Å.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manifestation of p-p Stacking Interactions in Luminescence Properties and Energy Transfer in Aromatically-Derived Tb, Eu and Gd Tris (pyrazolyl) borate Complexes
Mikhalyova EA, et al.
Inorganic Chemistry, 54(7), 3125-3133 (2015)
Structural, optical and electrical characteristics of transparent bismuth vanadate films deposited on indium tin oxide coated glass substrates
Kumari N, et al.
Journal of Materials Science: Materials in Electronics, 21(11), 1107-1114 (2010)
Thin polyphenylene vinylene electrophoretically and spin-coated films-photoelectrical properties
Mladenova D, et al.
Journal of Physics. Conference Series, 398(1) (2012)
Nanostructured iron (III) oxides: From design to gas-and liquid-phase photo-catalytic applications
Carraro G, et al.
Thin Solid Films, 564, 121-127 (2014)
Online coupling of digital microfluidic devices with mass spectrometry detection using an eductor with electrospray ionization.
Baker CA and Roper MG
Analytical Chemistry, 2955-2960 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service