Skip to Content
Merck
All Photos(1)

Key Documents

729086

Sigma-Aldrich

Poly(ethylene glycol) diacrylate

average Mn 1,000, acrylate, MEHQ as inhibitor (may contain)

Synonym(s):

Polyethylene glycol, PEG diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

Poly(ethylene glycol) diacrylate, average Mn 1,000, contains MEHQ as inhibitor

form

solid

Quality Level

mol wt

average Mn 1,000

contains

MEHQ as inhibitor
≤1,500 ppm MEHQ as inhibitor (may contain)

composition

C2H3C(O)(OC2H4)nOC(O)C2H3

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

transition temp

Tm 32-37 °C

Mw/Mn

≤1.1

Ω-end

acrylate

α-end

acrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

SMILES string

OCCO.OC(=O)C=C

InChI

1S/C8H10O4/c1-3-7(9)11-5-6-12-8(10)4-2/h3-4H,1-2,5-6H2

InChI key

KUDUQBURMYMBIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) diacrylate (PEGDA) is a polyethylene glycol (PEG) based material that is used as a prepolymer solution that can be used in the formation of a cross-linked polymeric system.

Application

PEGDA has a wide range of usage potentially ranging from tissue engineering, photonics and other biological applications.

Preparation Note

Synthesized with an initial concentration of ≤1,500 ppm MEHQ

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural colour printing using a magnetically tunable and lithographically fixable photonic crystal
Kim H, et al.
Nature Photonics, 3(9), 534-534 (2009)
Photoencapsulation of osteoblasts in injectable RGD-modified PEG hydrogels for bone tissue engineering
Burdick JA and Anseth KS
Biomaterials, 23(22), 4315-4323 (2002)
Preparation and characterization of crosslinked poly (ethylene glycol) diacrylate hydrogels as fouling-resistant membrane coating materials
Ju H, et al.
Journal of Membrane Science , 330(1-2), 180-188 (2009)
Release of protein from highly cross-linked hydrogels of poly (ethylene glycol) diacrylate fabricated by UV polymerization
Mellott MB, et al.
Biomaterials, 22(9), 929-941 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service