Skip to Content
Merck
All Photos(1)

Key Documents

725692

Sigma-Aldrich

Poly(ethylene glycol) dimethacrylate

average MN 20,000, cross-linking reagent polymerization reactions, methacrylate, ≤1, 500 ppm MEHQ as inhibitor (may contain)

Synonym(s):

Polyethylene glycol, PEG dimethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5C(O)(OCH2CH2)nOC(O)C3H5
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor

form

powder

mol wt

average Mn 20,000

contains

MEHQ as inhibitor
≤1,500 ppm MEHQ as inhibitor (may contain)

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

bp

>200 °C/2 mmHg (lit.)

transition temp

Tm 59-64 °C

Mw/Mn

≤1.1

Ω-end

methacrylate

α-end

methacrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

SMILES string

OCCO.CC(=C)C(O)=O

InChI

1S/C10H14O4/c1-7(2)9(11)13-5-6-14-10(12)8(3)4/h1,3,5-6H2,2,4H3

InChI key

STVZJERGLQHEKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Synthesized with an initial concentration of ≤1,500 ppm MEHQ

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuan Mu et al.
Lab on a chip, 13(8), 1612-1618 (2013-03-05)
Engineering functional vascular networks in vitro is critical for tissue engineering and a variety of applications. There is still a general lack of straightforward approaches for recapitulating specific structures and functions of vasculature. This report describes a microfluidic method that
C Aulin et al.
Laboratory animals, 47(1), 58-65 (2013-03-08)
Articular cartilage has a limited capacity for self-repair in adult humans, and methods used to stimulate regeneration often result in re-growth of fibrous cartilage, which has lower durability. No current treatment option can provide complete repair. The possibility of growth
Pelagie M Favi et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 1935-1944 (2013-03-19)
The culture of multipotent mesenchymal stem cells on natural biopolymers holds great promise for treatments of connective tissue disorders such as osteoarthritis. The safety and performance of such therapies relies on the systematic in vitro evaluation of the developed stem
Alyssa J Reiffel et al.
PloS one, 8(2), e56506-e56506 (2013-02-26)
Autologous techniques for the reconstruction of pediatric microtia often result in suboptimal aesthetic outcomes and morbidity at the costal cartilage donor site. We therefore sought to combine digital photogrammetry with CAD/CAM techniques to develop collagen type I hydrogel scaffolds and
Hailuo Fu et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 2245-2250 (2013-03-19)
Implants that simultaneously function as an osteoconductive matrix and as a device for local drug or growth factor delivery could provide an attractive system for bone regeneration. In our previous work, we prepared hollow hydroxyapatite (abbreviated HA) microspheres with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service