Skip to Content
Merck
All Photos(2)

Documents

701963

Sigma-Aldrich

Poly(ethylene glycol) diacrylate

average Mn 6,000, acrylate, ≤1,500 ppm MEHQ as inhibitor

Synonym(s):

Polyethylene glycol, PEG diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) diacrylate, average Mn 6,000, contains ≤1500 ppm MEHQ as inhibitor

form

solid

mol wt

average Mn 6,000

contains

≤1500 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

transition temp

Tm 59-63 °C

Ω-end

acrylate

α-end

acrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

SMILES string

OCCO.OC(=O)C=C

InChI

1S/C8H10O4/c1-3-7(9)11-5-6-12-8(10)4-2/h3-4H,1-2,5-6H2

InChI key

KUDUQBURMYMBIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol)diacrylate (PEGDA) is a long chain, hydrophilic and crosslinking monomer widelyused in tissue engineering.

Application

PEGDA is widely used as a scaffolding material for tissue engineering applications due to its biocompatibility and inherent resistance to protein adhesion.

It can be used as an alloying agent to prepare polymer membranes for gas separation applications. For example, an alloyed poly(Ether Block Amide)/ PEGDA membrane can be used for the separation of CO2/H2.

It can also be used as aprecursor to fabricate polymer electrolyte membranes(PEMs) for flexible Li-ionbatteries. The addition of PEGDA enhances the ionic conductivity, thermal stability,and mechanical toughness of PEMs.

Features and Benefits

  • Highly hydrophilic
  • Non-toxic
  • Biocompatible
  • Non-immunogenic

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaun P Garland et al.
Langmuir : the ACS journal of surfaces and colloids, 30(8), 2101-2108 (2014-02-15)
A growing body of literature broadly documents that a wide array of fundamental cell behaviors are modulated by the physical attributes of the cellular microenvironment, yet in vitro assays are typically carried out using tissue culture plastic or glass substrates
Eyal Karzbrun et al.
Nature physics, 14(5), 515-522 (2018-05-16)
Human brain wrinkling has been implicated in neurodevelopmental disorders and yet its origins remain unknown. Polymer gel models suggest that wrinkling emerges spontaneously due to compression forces arising during differential swelling, but these ideas have not been tested in a
Sandeep Ameta et al.
Nature communications, 12(1), 842-842 (2021-02-10)
Discovering autocatalytic chemistries that can evolve is a major goal in systems chemistry and a critical step towards understanding the origin of life. Autocatalytic networks have been discovered in various chemistries, but we lack a general understanding of how network
Adel Badria et al.
Journal of materials science. Materials in medicine, 29(11), 175-175 (2018-11-11)
Heart valve diseases remain common in industrialized countries. Bioprosthetic heart valves, introduced as free of anticoagulation therapy alternatives to mechanical substitutes. Still they suffer from long term failure due to calcification. Different treatment methods introduced to inhibit calcification, have so
Ruohong Shi et al.
Small (Weinheim an der Bergstrasse, Germany), 16(37), e2002946-e2002946 (2020-08-11)
Hydrogels with the ability to change shape in response to biochemical stimuli are important for biosensing, smart medicine, drug delivery, and soft robotics. Here, a family of multicomponent DNA polymerization motor gels with different polymer backbones is created, including acrylamide-co-bis-acrylamide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service